Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96661591-96661864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
UniGene
Hs.534025
Gene Chromosome Location
Chr.2: 96642719-96704887 on Build GRCh38
Transcripts
NM_001293083.1 XM_011512110.2 XM_011512111.2 XM_011512113.2 XM_011512114.2 XM_011512115.2 XM_011512116.2 XM_011512117.2 XM_011512118.2 XM_011512119.2 XM_011512120.2 XM_011512122.2 XM_011512125.2 XM_011512126.2 XM_011512127.2 XM_011512128.2 XM_017005222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1410069 c.870C>T p.T290T Chr.2: 96661694-96661694 on Build GRCh38
1023937 c.945C>T p.Y315Y Chr.2: 96661769-96661769 on Build GRCh38
1023938 c.948C>T p.C316C Chr.2: 96661772-96661772 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567379170 Chr.2: 96661732-96661732 on Build GRCh38
rs771529643 Chr.2: 96661747-96661747 on Build GRCh38
rs745956101 Chr.2: 96661819-96661819 on Build GRCh38
rs548462259 Chr.2: 96661703-96661703 on Build GRCh38
rs867002330 Chr.2: 96661773-96661773 on Build GRCh38
rs1004391958 Chr.2: 96661687-96661687 on Build GRCh38
rs766921663 Chr.2: 96661672-96661672 on Build GRCh38
rs966646027 Chr.2: 96661617-96661617 on Build GRCh38
rs930998744 Chr.2: 96661632-96661632 on Build GRCh38
rs758668608 Chr.2: 96661733-96661733 on Build GRCh38
rs780144758 Chr.2: 96661695-96661695 on Build GRCh38
rs368115806 Chr.2: 96661706-96661706 on Build GRCh38
rs996658100 Chr.2: 96661701-96661701 on Build GRCh38
rs768814730 Chr.2: 96661726-96661726 on Build GRCh38
rs773098785 Chr.2: 96661839-96661839 on Build GRCh38
rs769673265 Chr.2: 96661776-96661776 on Build GRCh38
rs769935357 Chr.2: 96661781-96661781 on Build GRCh38
rs529697801 Chr.2: 96661760-96661760 on Build GRCh38
rs1010815886 Chr.2: 96661718-96661718 on Build GRCh38
rs975983406 Chr.2: 96661772-96661772 on Build GRCh38
rs747468079 Chr.2: 96661735-96661735 on Build GRCh38
rs1032362119 Chr.2: 96661649-96661649 on Build GRCh38
rs919193639 Chr.2: 96661816-96661816 on Build GRCh38
rs954157949 Chr.2: 96661683-96661683 on Build GRCh38
rs757282875 Chr.2: 96661671-96661671 on Build GRCh38
rs749485474 Chr.2: 96661725-96661725 on Build GRCh38
rs908943151 Chr.2: 96661840-96661840 on Build GRCh38
rs751419176 Chr.2: 96661644-96661644 on Build GRCh38
rs138327933 Chr.2: 96661631-96661632 on Build GRCh38
rs761428331 Chr.2: 96661727-96661727 on Build GRCh38
rs778242619 Chr.2: 96661734-96661734 on Build GRCh38
rs1022084370 Chr.2: 96661724-96661724 on Build GRCh38
rs531968420 Chr.2: 96661694-96661694 on Build GRCh38
rs267599499 Chr.2: 96661693-96661693 on Build GRCh38
rs36121442 Chr.2: 96661614-96661615 on Build GRCh38

View Full Product Details