Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3493801-3494074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOHH
Entrez Gene ID
83475
Gene Name
deoxyhypusine hydroxylase/monooxygenase
Gene Aliases
HLRC1, hDOHH
UniGene
Hs.515064
Gene Chromosome Location
Chr.19: 3490821-3500940 on Build GRCh38
Transcripts
NM_001145165.1 NM_031304.4 XM_011528341.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357112 c.306A>G p.E102E Chr.19: 3494073-3494073 on Build GRCh38
319982 c.349G>T p.E117* Chr.19: 3494030-3494030 on Build GRCh38
1284604 c.324G>A p.K108K Chr.19: 3494055-3494055 on Build GRCh38
4488462 c.332C>T p.S111F Chr.19: 3494047-3494047 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563883288 Chr.19: 3493909-3493909 on Build GRCh38
rs1054692968 Chr.19: 3493866-3493866 on Build GRCh38
rs375620572 Chr.19: 3494017-3494017 on Build GRCh38
rs770266870 Chr.19: 3494018-3494018 on Build GRCh38
rs962953863 Chr.19: 3493839-3493839 on Build GRCh38
rs750207414 Chr.19: 3494005-3494005 on Build GRCh38
rs758720511 Chr.19: 3494006-3494006 on Build GRCh38
rs926178386 Chr.19: 3493972-3493972 on Build GRCh38
rs369076634 Chr.19: 3494043-3494043 on Build GRCh38
rs771254943 Chr.19: 3494030-3494030 on Build GRCh38
rs199800462 Chr.19: 3494033-3494033 on Build GRCh38
rs886859460 Chr.19: 3494028-3494028 on Build GRCh38
rs911998540 Chr.19: 3493912-3493912 on Build GRCh38
rs918349597 Chr.19: 3493822-3493822 on Build GRCh38
rs763657849 Chr.19: 3493992-3493992 on Build GRCh38
rs753778810 Chr.19: 3493993-3493993 on Build GRCh38
rs778579587 Chr.19: 3494003-3494003 on Build GRCh38
rs757164047 Chr.19: 3494002-3494002 on Build GRCh38
rs191602512 Chr.19: 3493926-3493926 on Build GRCh38
rs11551227 Chr.19: 3494037-3494037 on Build GRCh38
rs927475348 Chr.19: 3493864-3493864 on Build GRCh38
rs750366983 Chr.19: 3494047-3494047 on Build GRCh38
rs569448679 Chr.19: 3493959-3493959 on Build GRCh38
rs765177422 Chr.19: 3494044-3494044 on Build GRCh38
rs747198722 Chr.19: 3494011-3494011 on Build GRCh38
rs549606329 Chr.19: 3493928-3493928 on Build GRCh38
rs1007112727 Chr.19: 3493997-3493997 on Build GRCh38
rs200540157 Chr.19: 3494014-3494014 on Build GRCh38
rs953436943 Chr.19: 3493888-3493888 on Build GRCh38
rs368267577 Chr.19: 3494032-3494032 on Build GRCh38
rs781316025 Chr.19: 3494016-3494016 on Build GRCh38
rs1041917295 Chr.19: 3493945-3493945 on Build GRCh38
rs760218452 Chr.19: 3493985-3493985 on Build GRCh38
rs921912021 Chr.19: 3493886-3493886 on Build GRCh38
rs975396723 Chr.19: 3493865-3493865 on Build GRCh38
rs1046361292 Chr.19: 3493840-3493840 on Build GRCh38
rs987527280 Chr.19: 3493893-3493893 on Build GRCh38
rs773597811 Chr.19: 3494019-3494019 on Build GRCh38
rs1041666264 Chr.19: 3493887-3493887 on Build GRCh38
rs534573908 Chr.19: 3494039-3494039 on Build GRCh38
rs780469210 Chr.19: 3494009-3494009 on Build GRCh38
rs749604084 Chr.19: 3494023-3494023 on Build GRCh38
rs893363927 Chr.19: 3493870-3493870 on Build GRCh38
rs949793665 Chr.19: 3493823-3493823 on Build GRCh38

View Full Product Details