Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99188201-99188436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA7
Entrez Gene ID
402569
Gene Name
karyopherin subunit alpha 7
Gene Aliases
IPOA8
UniGene
Hs.253050
Gene Chromosome Location
Chr.7: 99145467-99219417 on Build GRCh38
Transcripts
XM_011516215.2 XM_017012208.1 XM_017012209.1 XM_017012210.1 XM_017012211.1 XM_017012212.1 XM_017012213.1 XM_017012214.1 XM_017012215.1 XM_017012216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3833594 c.873C>A p.L291L Chr.7: 99188327-99188327 on Build GRCh38
5551958 c.810C>T p.D270D Chr.7: 99188390-99188390 on Build GRCh38
3431929 c.848G>C p.G283A Chr.7: 99188352-99188352 on Build GRCh38
6861788 c.869T>G p.V290G Chr.7: 99188331-99188331 on Build GRCh38
3995770 c.896T>C p.V299A Chr.7: 99188304-99188304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041727630 Chr.7: 99188287-99188287 on Build GRCh38
rs764195081 Chr.7: 99188251-99188251 on Build GRCh38
rs755041223 Chr.7: 99188321-99188321 on Build GRCh38
rs745528364 Chr.7: 99188374-99188374 on Build GRCh38
rs191355534 Chr.7: 99188335-99188335 on Build GRCh38
rs369219037 Chr.7: 99188317-99188317 on Build GRCh38
rs887719282 Chr.7: 99188385-99188385 on Build GRCh38
rs1023709548 Chr.7: 99188415-99188415 on Build GRCh38
rs777728498 Chr.7: 99188257-99188257 on Build GRCh38
rs771282512 Chr.7: 99188391-99188392 on Build GRCh38
rs183724463 Chr.7: 99188389-99188389 on Build GRCh38
rs796635090 Chr.7: 99188224-99188224 on Build GRCh38
rs747222694 Chr.7: 99188332-99188332 on Build GRCh38
rs960847629 Chr.7: 99188401-99188401 on Build GRCh38
rs916564770 Chr.7: 99188253-99188253 on Build GRCh38
rs747351392 Chr.7: 99188255-99188256 on Build GRCh38
rs778588208 Chr.7: 99188372-99188372 on Build GRCh38
rs764968209 Chr.7: 99188399-99188399 on Build GRCh38
rs944290941 Chr.7: 99188263-99188263 on Build GRCh38
rs767510250 Chr.7: 99188333-99188333 on Build GRCh38
rs116113870 Chr.7: 99188262-99188262 on Build GRCh38
rs935460857 Chr.7: 99188270-99188270 on Build GRCh38
rs761633120 Chr.7: 99188398-99188398 on Build GRCh38
rs948095877 Chr.7: 99188259-99188259 on Build GRCh38
rs774236036 Chr.7: 99188231-99188231 on Build GRCh38
rs377664445 Chr.7: 99188376-99188376 on Build GRCh38
rs528934094 Chr.7: 99188350-99188350 on Build GRCh38
rs749626280 Chr.7: 99188354-99188354 on Build GRCh38
rs756982898 Chr.7: 99188414-99188414 on Build GRCh38
rs774578304 Chr.7: 99188405-99188405 on Build GRCh38
rs900159181 Chr.7: 99188371-99188371 on Build GRCh38
rs572557822 Chr.7: 99188292-99188292 on Build GRCh38
rs371196094 Chr.7: 99188307-99188307 on Build GRCh38
rs550198713 Chr.7: 99188355-99188355 on Build GRCh38
rs569828674 Chr.7: 99188268-99188268 on Build GRCh38
rs776749940 Chr.7: 99188393-99188393 on Build GRCh38
rs369700638 Chr.7: 99188390-99188390 on Build GRCh38
rs200524486 Chr.7: 99188377-99188377 on Build GRCh38
rs561494283 Chr.7: 99188345-99188345 on Build GRCh38

View Full Product Details