Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24632864-24633135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMB
Entrez Gene ID
3002
Gene Name
granzyme B
Gene Aliases
CCPI, CGL-1, CGL1, CSP-B, CSPB, CTLA1, CTSGL1, HLP, SECT
UniGene
Hs.1051
Gene Chromosome Location
Chr.14: 24630954-24634226 on Build GRCh38
Transcripts
NM_004131.4 XM_011536685.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4050350 c.85C>G p.P29A Chr.14: 24633033-24633033 on Build GRCh38
6849182 c.148G>A p.G50S Chr.14: 24632970-24632970 on Build GRCh38
6538061 c.250G>C p.G84R Chr.14: 24632970-24632970 on Build GRCh38
3706228 c.125A>G p.D42G Chr.14: 24632993-24632993 on Build GRCh38
4050351 c.187C>G p.P63A Chr.14: 24633033-24633033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912310282 Chr.14: 24633020-24633020 on Build GRCh38
rs1039263374 Chr.14: 24632888-24632888 on Build GRCh38
rs781125360 Chr.14: 24633114-24633114 on Build GRCh38
rs768588540 Chr.14: 24633024-24633024 on Build GRCh38
rs564414889 Chr.14: 24632970-24632970 on Build GRCh38
rs770449395 Chr.14: 24632914-24632914 on Build GRCh38
rs759571695 Chr.14: 24633042-24633042 on Build GRCh38
rs986579745 Chr.14: 24633008-24633008 on Build GRCh38
rs139103546 Chr.14: 24633052-24633052 on Build GRCh38
rs763072582 Chr.14: 24632949-24632949 on Build GRCh38
rs200381701 Chr.14: 24632972-24632972 on Build GRCh38
rs988057545 Chr.14: 24633066-24633066 on Build GRCh38
rs769831969 Chr.14: 24632943-24632943 on Build GRCh38
rs923986255 Chr.14: 24633029-24633029 on Build GRCh38
rs757747585 Chr.14: 24633062-24633062 on Build GRCh38
rs756689392 Chr.14: 24632975-24632975 on Build GRCh38
rs778477049 Chr.14: 24632928-24632928 on Build GRCh38
rs775333412 Chr.14: 24632941-24632941 on Build GRCh38
rs199546734 Chr.14: 24633002-24633002 on Build GRCh38
rs111296281 Chr.14: 24632940-24632940 on Build GRCh38
rs766574069 Chr.14: 24632950-24632950 on Build GRCh38
rs754267669 Chr.14: 24633058-24633058 on Build GRCh38
rs769914418 Chr.14: 24633035-24633035 on Build GRCh38
rs766594028 Chr.14: 24632979-24632979 on Build GRCh38
rs771725823 Chr.14: 24633022-24633022 on Build GRCh38
rs372092724 Chr.14: 24633068-24633068 on Build GRCh38
rs767677921 Chr.14: 24632967-24632967 on Build GRCh38
rs968157813 Chr.14: 24633012-24633012 on Build GRCh38
rs761842669 Chr.14: 24633026-24633026 on Build GRCh38
rs779606646 Chr.14: 24633023-24633023 on Build GRCh38
rs374673076 Chr.14: 24632994-24632994 on Build GRCh38
rs774464415 Chr.14: 24633040-24633040 on Build GRCh38
rs776646060 Chr.14: 24633025-24633025 on Build GRCh38
rs748242826 Chr.14: 24632895-24632895 on Build GRCh38
rs141950099 Chr.14: 24632971-24632971 on Build GRCh38
rs771688554 Chr.14: 24632938-24632938 on Build GRCh38
rs150732480 Chr.14: 24633028-24633028 on Build GRCh38
rs746648398 Chr.14: 24633110-24633110 on Build GRCh38
rs756619054 Chr.14: 24632898-24632898 on Build GRCh38
rs944609648 Chr.14: 24633102-24633102 on Build GRCh38
rs776733343 Chr.14: 24632946-24632946 on Build GRCh38
rs767736704 Chr.14: 24633047-24633047 on Build GRCh38
rs748753120 Chr.14: 24632910-24632910 on Build GRCh38
rs140998431 Chr.14: 24633097-24633097 on Build GRCh38
rs759793850 Chr.14: 24632964-24632964 on Build GRCh38
rs8192917 Chr.14: 24632954-24632954 on Build GRCh38
rs768676395 Chr.14: 24632945-24632945 on Build GRCh38
rs371428669 Chr.14: 24632909-24632909 on Build GRCh38
rs745539169 Chr.14: 24632935-24632935 on Build GRCh38
rs747395753 Chr.14: 24633064-24633064 on Build GRCh38
rs749909360 Chr.14: 24633093-24633093 on Build GRCh38
rs779700655 Chr.14: 24633099-24633099 on Build GRCh38
rs368712815 Chr.14: 24633051-24633051 on Build GRCh38

View Full Product Details