Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101831051-101831290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP2
Entrez Gene ID
30819
Gene Name
potassium voltage-gated channel interacting protein 2
Gene Aliases
KCHIP2
UniGene
Hs.97044
Gene Chromosome Location
Chr.10: 101825971-101843920 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5954974 c.133T>A p.C45S Chr.10: 101831108-101831108 on Build GRCh38
7321254 c.165T>C p.S55S Chr.10: 101831076-101831076 on Build GRCh38
914075 c.141G>A p.G47G Chr.10: 101831100-101831100 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748206100 Chr.10: 101831199-101831199 on Build GRCh38
rs946551220 Chr.10: 101831076-101831079 on Build GRCh38
rs745729177 Chr.10: 101831156-101831156 on Build GRCh38
rs762181863 Chr.10: 101831089-101831089 on Build GRCh38
rs772385248 Chr.10: 101831204-101831204 on Build GRCh38
rs955311334 Chr.10: 101831270-101831270 on Build GRCh38
rs766718618 Chr.10: 101831117-101831117 on Build GRCh38
rs575471302 Chr.10: 101831159-101831159 on Build GRCh38
rs780682184 Chr.10: 101831154-101831154 on Build GRCh38
rs749249006 Chr.10: 101831183-101831183 on Build GRCh38
rs372224281 Chr.10: 101831103-101831103 on Build GRCh38
rs1016746458 Chr.10: 101831179-101831179 on Build GRCh38
rs1043648276 Chr.10: 101831174-101831175 on Build GRCh38
rs767627046 Chr.10: 101831097-101831097 on Build GRCh38
rs546171339 Chr.10: 101831237-101831237 on Build GRCh38
rs745434445 Chr.10: 101831103-101831113 on Build GRCh38
rs1034733270 Chr.10: 101831268-101831268 on Build GRCh38
rs751058723 Chr.10: 101831150-101831150 on Build GRCh38
rs755413672 Chr.10: 101831128-101831128 on Build GRCh38
rs999592449 Chr.10: 101831213-101831213 on Build GRCh38
rs201061453 Chr.10: 101831209-101831209 on Build GRCh38
rs757004027 Chr.10: 101831153-101831153 on Build GRCh38
rs773664622 Chr.10: 101831100-101831100 on Build GRCh38
rs779923232 Chr.10: 101831177-101831177 on Build GRCh38
rs754374069 Chr.10: 101831120-101831120 on Build GRCh38
rs763609219 Chr.10: 101831139-101831139 on Build GRCh38
rs774638596 Chr.10: 101831197-101831197 on Build GRCh38
rs72552059 Chr.10: 101831165-101831165 on Build GRCh38
rs768698758 Chr.10: 101831195-101831195 on Build GRCh38

View Full Product Details