Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57198238-57198463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HESX1
Entrez Gene ID
8820
Gene Name
HESX homeobox 1
Gene Aliases
ANF, CPHD5, RPX
UniGene
Hs.171980
Gene Chromosome Location
Chr.3: 57197838-57227643 on Build GRCh38
Transcripts
NM_003865.2 XM_005265526.4 XM_006713379.3 XM_011534204.2 XM_011534205.2 XM_017007421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338318 c.476G>A p.R159Q Chr.3: 57198279-57198279 on Build GRCh38
1424825 c.479G>A p.R160H Chr.3: 57198276-57198276 on Build GRCh38
6327061 c.479G>T p.R160L Chr.3: 57198276-57198276 on Build GRCh38
4119527 c.421T>A p.L141I Chr.3: 57198429-57198429 on Build GRCh38
398237 c.500A>G p.H167R Chr.3: 57198255-57198255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757267705 Chr.3: 57198395-57198396 on Build GRCh38
rs1012304897 Chr.3: 57198354-57198354 on Build GRCh38
rs587776664 Chr.3: 57198399-57198400 on Build GRCh38
rs753824888 Chr.3: 57198365-57198365 on Build GRCh38
rs376071780 Chr.3: 57198373-57198373 on Build GRCh38
rs894964698 Chr.3: 57198353-57198353 on Build GRCh38
rs778799278 Chr.3: 57198428-57198429 on Build GRCh38
rs773149249 Chr.3: 57198273-57198273 on Build GRCh38
rs1019646173 Chr.3: 57198341-57198341 on Build GRCh38
rs112703580 Chr.3: 57198298-57198298 on Build GRCh38
rs764058535 Chr.3: 57198369-57198369 on Build GRCh38
rs770886420 Chr.3: 57198280-57198280 on Build GRCh38
rs760082717 Chr.3: 57198344-57198346 on Build GRCh38
rs530696048 Chr.3: 57198332-57198332 on Build GRCh38
rs28936702 Chr.3: 57198277-57198277 on Build GRCh38
rs104893742 Chr.3: 57198405-57198405 on Build GRCh38
rs763758902 Chr.3: 57198363-57198363 on Build GRCh38
rs1000783269 Chr.3: 57198304-57198304 on Build GRCh38
rs542387569 Chr.3: 57198316-57198316 on Build GRCh38
rs760503914 Chr.3: 57198274-57198274 on Build GRCh38
rs987783507 Chr.3: 57198337-57198337 on Build GRCh38
rs766234350 Chr.3: 57198276-57198276 on Build GRCh38

View Full Product Details