Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218134922-218135183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR2
Entrez Gene ID
3579
Gene Name
C-X-C motif chemokine receptor 2
Gene Aliases
CD182, CDw128b, CMKAR2, IL8R2, IL8RA, IL8RB
UniGene
Hs.846
Gene Chromosome Location
Chr.2: 218125290-218137253 on Build GRCh38
Transcripts
NM_001168298.1 NM_001557.3 XM_005246530.3 XM_017003990.1 XM_017003991.1 XM_017003992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764840 c.137T>C p.I46T Chr.2: 218134938-218134938 on Build GRCh38
3043197 c.244G>A p.V82I Chr.2: 218135045-218135045 on Build GRCh38
1016299 c.236G>A p.G79D Chr.2: 218135037-218135037 on Build GRCh38
4951818 c.233T>A p.V78D Chr.2: 218135034-218135034 on Build GRCh38
1016298 c.234C>T p.V78V Chr.2: 218135035-218135035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759645813 Chr.2: 218135082-218135082 on Build GRCh38
rs199535002 Chr.2: 218135073-218135073 on Build GRCh38
rs199633255 Chr.2: 218135024-218135024 on Build GRCh38
rs368269858 Chr.2: 218135006-218135006 on Build GRCh38
rs200115845 Chr.2: 218135120-218135120 on Build GRCh38
rs150483744 Chr.2: 218135035-218135035 on Build GRCh38
rs867311007 Chr.2: 218134954-218134954 on Build GRCh38
rs753356883 Chr.2: 218135154-218135154 on Build GRCh38
rs762103956 Chr.2: 218134991-218134991 on Build GRCh38
rs779316036 Chr.2: 218135069-218135069 on Build GRCh38
rs983718457 Chr.2: 218135128-218135128 on Build GRCh38
rs756891890 Chr.2: 218134947-218134947 on Build GRCh38
rs534692959 Chr.2: 218134990-218134990 on Build GRCh38
rs11574748 Chr.2: 218134982-218134982 on Build GRCh38
rs776644645 Chr.2: 218135081-218135081 on Build GRCh38
rs780729371 Chr.2: 218134949-218134949 on Build GRCh38
rs201754155 Chr.2: 218135045-218135045 on Build GRCh38
rs138773569 Chr.2: 218135039-218135039 on Build GRCh38
rs201513058 Chr.2: 218135114-218135114 on Build GRCh38
rs779873701 Chr.2: 218135150-218135150 on Build GRCh38
rs201294973 Chr.2: 218135005-218135005 on Build GRCh38
rs773306755 Chr.2: 218134995-218134995 on Build GRCh38
rs199770538 Chr.2: 218134965-218134965 on Build GRCh38
rs767734877 Chr.2: 218134992-218134992 on Build GRCh38
rs760780846 Chr.2: 218135077-218135077 on Build GRCh38
rs769442813 Chr.2: 218134962-218134962 on Build GRCh38
rs780840157 Chr.2: 218135040-218135040 on Build GRCh38
rs752383293 Chr.2: 218135072-218135072 on Build GRCh38
rs761868394 Chr.2: 218135116-218135116 on Build GRCh38
rs1014647889 Chr.2: 218135117-218135117 on Build GRCh38
rs769324569 Chr.2: 218134983-218134983 on Build GRCh38
rs921960299 Chr.2: 218134975-218134975 on Build GRCh38
rs763954636 Chr.2: 218135101-218135101 on Build GRCh38
rs753962612 Chr.2: 218135026-218135026 on Build GRCh38
rs900536187 Chr.2: 218135063-218135063 on Build GRCh38
rs372364370 Chr.2: 218135044-218135044 on Build GRCh38
rs745318598 Chr.2: 218134959-218134959 on Build GRCh38
rs935179968 Chr.2: 218134971-218134971 on Build GRCh38
rs771195420 Chr.2: 218135080-218135080 on Build GRCh38
rs149364972 Chr.2: 218135036-218135036 on Build GRCh38
rs200819332 Chr.2: 218135071-218135071 on Build GRCh38
rs751466333 Chr.2: 218135113-218135113 on Build GRCh38
rs963559162 Chr.2: 218135151-218135151 on Build GRCh38
rs1051685752 Chr.2: 218134998-218134998 on Build GRCh38
rs1054064754 Chr.2: 218135014-218135014 on Build GRCh38
rs750271295 Chr.2: 218135145-218135145 on Build GRCh38
rs142747099 Chr.2: 218135075-218135075 on Build GRCh38
rs767211869 Chr.2: 218135118-218135118 on Build GRCh38
rs147368052 Chr.2: 218135149-218135149 on Build GRCh38
rs769234467 Chr.2: 218135062-218135062 on Build GRCh38

View Full Product Details