Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70454129-70454402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG3
Entrez Gene ID
1741
Gene Name
discs large MAGUK scaffold protein 3
Gene Aliases
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
UniGene
Hs.721586
Gene Chromosome Location
Chr.X: 70444850-70505490 on Build GRCh38
Transcripts
NM_020730.2 NM_021120.3 XM_005262248.3 XM_006724625.2 XM_006724626.2 XM_011530883.1 XM_017029322.1 XM_017029323.1 XM_017029324.1 NR_046586.1 NR_109801.1

Gene Symbol
DLG3-AS1
Entrez Gene ID
100873930
Gene Name
DLG3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.571699
Gene Chromosome Location
Chr.X: 70452956-70455994 on Build GRCh38
Transcripts
NM_020730.2 NM_021120.3 XM_005262248.3 XM_006724625.2 XM_006724626.2 XM_011530883.1 XM_017029322.1 XM_017029323.1 XM_017029324.1 NR_046586.1 NR_109801.1

SNP Detail

SNP ID Chromosome Location
rs761856095 Chr.X: 70454283-70454283 on Build GRCh38
rs1010299412 Chr.X: 70454362-70454362 on Build GRCh38
rs374740363 Chr.X: 70454313-70454313 on Build GRCh38
rs917016084 Chr.X: 70454161-70454161 on Build GRCh38
rs768558801 Chr.X: 70454277-70454277 on Build GRCh38
rs749085371 Chr.X: 70454272-70454272 on Build GRCh38
rs368606425 Chr.X: 70454178-70454178 on Build GRCh38
rs797045525 Chr.X: 70454321-70454323 on Build GRCh38
rs754183509 Chr.X: 70454349-70454349 on Build GRCh38
rs750662881 Chr.X: 70454192-70454192 on Build GRCh38
rs942541567 Chr.X: 70454262-70454262 on Build GRCh38
rs767324184 Chr.X: 70454296-70454296 on Build GRCh38
rs780218523 Chr.X: 70454362-70454366 on Build GRCh38
rs766716994 Chr.X: 70454332-70454332 on Build GRCh38
rs375285809 Chr.X: 70454176-70454176 on Build GRCh38
rs769457554 Chr.X: 70454271-70454271 on Build GRCh38
rs1038647104 Chr.X: 70454326-70454326 on Build GRCh38
rs755307053 Chr.X: 70454167-70454167 on Build GRCh38
rs745344573 Chr.X: 70454242-70454242 on Build GRCh38
rs371545673 Chr.X: 70454299-70454299 on Build GRCh38
rs398122847 Chr.X: 70454284-70454284 on Build GRCh38
rs892966006 Chr.X: 70454331-70454331 on Build GRCh38

View Full Product Details