Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2932476-2932725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD10
Entrez Gene ID
10227
Gene Name
major facilitator superfamily domain containing 10
Gene Aliases
TETRAN, TETTRAN
UniGene
Hs.632581
Gene Chromosome Location
Chr.4: 2930560-2934859 on Build GRCh38
Transcripts
NM_001120.4 NM_001146069.1 XM_005247927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836279 c.531+1G>C p.? Chr.4: 2932599-2932599 on Build GRCh38
261991 c.518G>A p.R173H Chr.4: 2932613-2932613 on Build GRCh38
305926 c.532-5C>T p.? Chr.4: 2932517-2932517 on Build GRCh38
5355980 c.423G>A p.A141A Chr.4: 2932708-2932708 on Build GRCh38
3825750 c.526G>T p.G176C Chr.4: 2932605-2932605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1263349 Chr.4: 2932581-2932581 on Build GRCh38
rs762542635 Chr.4: 2932658-2932658 on Build GRCh38
rs748949364 Chr.4: 2932642-2932651 on Build GRCh38
rs765651773 Chr.4: 2932692-2932692 on Build GRCh38
rs746876932 Chr.4: 2932592-2932592 on Build GRCh38
rs748533846 Chr.4: 2932496-2932496 on Build GRCh38
rs764492126 Chr.4: 2932668-2932668 on Build GRCh38
rs769478098 Chr.4: 2932654-2932654 on Build GRCh38
rs761308208 Chr.4: 2932663-2932663 on Build GRCh38
rs766548882 Chr.4: 2932705-2932705 on Build GRCh38
rs909508524 Chr.4: 2932508-2932508 on Build GRCh38
rs199847211 Chr.4: 2932601-2932601 on Build GRCh38
rs776223295 Chr.4: 2932599-2932599 on Build GRCh38
rs745801383 Chr.4: 2932511-2932511 on Build GRCh38
rs769424992 Chr.4: 2932604-2932604 on Build GRCh38
rs755592589 Chr.4: 2932588-2932588 on Build GRCh38
rs777951326 Chr.4: 2932554-2932554 on Build GRCh38
rs762235084 Chr.4: 2932691-2932691 on Build GRCh38
rs750636561 Chr.4: 2932628-2932628 on Build GRCh38
rs145796300 Chr.4: 2932648-2932648 on Build GRCh38
rs377540995 Chr.4: 2932642-2932642 on Build GRCh38
rs767991561 Chr.4: 2932542-2932542 on Build GRCh38
rs201068903 Chr.4: 2932613-2932613 on Build GRCh38
rs764520359 Chr.4: 2932616-2932616 on Build GRCh38
rs780851136 Chr.4: 2932498-2932498 on Build GRCh38
rs946953148 Chr.4: 2932690-2932690 on Build GRCh38
rs780074847 Chr.4: 2932587-2932587 on Build GRCh38
rs369595481 Chr.4: 2932519-2932519 on Build GRCh38
rs778023916 Chr.4: 2932500-2932500 on Build GRCh38
rs750689852 Chr.4: 2932697-2932697 on Build GRCh38
rs972216433 Chr.4: 2932699-2932699 on Build GRCh38
rs778330576 Chr.4: 2932625-2932625 on Build GRCh38
rs754144124 Chr.4: 2932624-2932624 on Build GRCh38
rs747918438 Chr.4: 2932651-2932651 on Build GRCh38
rs772760097 Chr.4: 2932611-2932611 on Build GRCh38
rs138530490 Chr.4: 2932708-2932708 on Build GRCh38
rs865940469 Chr.4: 2932557-2932557 on Build GRCh38
rs763759416 Chr.4: 2932528-2932528 on Build GRCh38
rs749431099 Chr.4: 2932509-2932509 on Build GRCh38
rs1027589631 Chr.4: 2932565-2932565 on Build GRCh38
rs754102820 Chr.4: 2932566-2932566 on Build GRCh38
rs755606930 Chr.4: 2932562-2932564 on Build GRCh38
rs770489232 Chr.4: 2932662-2932662 on Build GRCh38
rs201365504 Chr.4: 2932510-2932510 on Build GRCh38
rs568902215 Chr.4: 2932571-2932572 on Build GRCh38
rs779197683 Chr.4: 2932573-2932573 on Build GRCh38
rs760095600 Chr.4: 2932534-2932534 on Build GRCh38
rs768513728 Chr.4: 2932597-2932597 on Build GRCh38
rs772182348 Chr.4: 2932582-2932582 on Build GRCh38
rs777181716 Chr.4: 2932676-2932676 on Build GRCh38
rs370161671 Chr.4: 2932649-2932649 on Build GRCh38
rs753041464 Chr.4: 2932545-2932545 on Build GRCh38
rs758630557 Chr.4: 2932702-2932702 on Build GRCh38
rs779944420 Chr.4: 2932646-2932646 on Build GRCh38
rs953440333 Chr.4: 2932596-2932596 on Build GRCh38
rs372224623 Chr.4: 2932573-2932574 on Build GRCh38
rs760490729 Chr.4: 2932520-2932520 on Build GRCh38
rs374576875 Chr.4: 2932641-2932641 on Build GRCh38
rs756339156 Chr.4: 2932499-2932499 on Build GRCh38
rs202141771 Chr.4: 2932614-2932614 on Build GRCh38
rs764731627 Chr.4: 2932531-2932531 on Build GRCh38
rs749828506 Chr.4: 2932657-2932657 on Build GRCh38
rs868603424 Chr.4: 2932517-2932517 on Build GRCh38
rs199921380 Chr.4: 2932530-2932530 on Build GRCh38
rs372985495 Chr.4: 2932568-2932568 on Build GRCh38
rs756530930 Chr.4: 2932551-2932551 on Build GRCh38

View Full Product Details