Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41083971-41084244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFYA
Entrez Gene ID
4800
Gene Name
nuclear transcription factor Y subunit alpha
Gene Aliases
CBF-A, CBF-B, HAP2, NF-YA
UniGene
Hs.10441
Gene Chromosome Location
Chr.6: 41072968-41102407 on Build GRCh38
Transcripts
NM_002505.4 NM_021705.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4937925 c.210C>G p.I70M Chr.6: 41084093-41084093 on Build GRCh38
6201685 c.234G>A p.M78I Chr.6: 41084117-41084117 on Build GRCh38
1444466 c.205C>A p.L69I Chr.6: 41084088-41084088 on Build GRCh38
1312189 c.215C>T p.S72L Chr.6: 41084098-41084098 on Build GRCh38
3627159 c.167A>T p.Q56L Chr.6: 41084050-41084050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020519086 Chr.6: 41084109-41084109 on Build GRCh38
rs572740457 Chr.6: 41084205-41084205 on Build GRCh38
rs200299860 Chr.6: 41084134-41084134 on Build GRCh38
rs768833544 Chr.6: 41084212-41084212 on Build GRCh38
rs372617775 Chr.6: 41084129-41084129 on Build GRCh38
rs754482435 Chr.6: 41084164-41084164 on Build GRCh38
rs775649217 Chr.6: 41084030-41084030 on Build GRCh38
rs767187310 Chr.6: 41084132-41084132 on Build GRCh38
rs576657046 Chr.6: 41084078-41084078 on Build GRCh38
rs957521469 Chr.6: 41084007-41084007 on Build GRCh38
rs780702130 Chr.6: 41084206-41084206 on Build GRCh38
rs201057112 Chr.6: 41084162-41084162 on Build GRCh38
rs746317318 Chr.6: 41084026-41084026 on Build GRCh38
rs763271966 Chr.6: 41084054-41084054 on Build GRCh38
rs773418404 Chr.6: 41084154-41084154 on Build GRCh38
rs749435561 Chr.6: 41084211-41084211 on Build GRCh38
rs774699992 Chr.6: 41084102-41084102 on Build GRCh38
rs868662543 Chr.6: 41084104-41084104 on Build GRCh38
rs760481342 Chr.6: 41084151-41084151 on Build GRCh38
rs781741048 Chr.6: 41084023-41084023 on Build GRCh38
rs750408038 Chr.6: 41084141-41084141 on Build GRCh38
rs558998840 Chr.6: 41084203-41084203 on Build GRCh38
rs781358022 Chr.6: 41084201-41084201 on Build GRCh38
rs17853896 Chr.6: 41084138-41084138 on Build GRCh38
rs772333591 Chr.6: 41084011-41084011 on Build GRCh38
rs538975038 Chr.6: 41084186-41084186 on Build GRCh38
rs757669414 Chr.6: 41084193-41084193 on Build GRCh38
rs769066664 Chr.6: 41084088-41084088 on Build GRCh38
rs753283289 Chr.6: 41084160-41084160 on Build GRCh38
rs374886963 Chr.6: 41084171-41084171 on Build GRCh38
rs140705580 Chr.6: 41084001-41084001 on Build GRCh38
rs764118250 Chr.6: 41084005-41084012 on Build GRCh38

View Full Product Details