Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186271552-186271825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11
Entrez Gene ID
2160
Gene Name
coagulation factor XI
Gene Aliases
FXI
UniGene
Hs.1430
Gene Chromosome Location
Chr.4: 186265945-186290727 on Build GRCh38
Transcripts
NM_000128.3 XM_005262821.3 XM_005262822.3 XM_005262823.3 XM_006714137.2 XM_017007884.1 XM_017007885.1 XM_017007886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584977 c.206A>G p.D69G Chr.4: 186271759-186271759 on Build GRCh38
4584976 c.206A>G p.D69G Chr.4: 186271759-186271759 on Build GRCh38
5429370 c.194C>T p.S65L Chr.4: 186271747-186271747 on Build GRCh38
4682177 c.118A>G p.T40A Chr.4: 186271671-186271671 on Build GRCh38
6099768 c.127G>T p.A43S Chr.4: 186271680-186271680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370826424 Chr.4: 186271658-186271658 on Build GRCh38
rs281875252 Chr.4: 186271705-186271705 on Build GRCh38
rs750564001 Chr.4: 186271790-186271790 on Build GRCh38
rs765301008 Chr.4: 186271789-186271789 on Build GRCh38
rs762978812 Chr.4: 186271757-186271757 on Build GRCh38
rs760244383 Chr.4: 186271618-186271618 on Build GRCh38
rs775977740 Chr.4: 186271636-186271636 on Build GRCh38
rs923326047 Chr.4: 186271700-186271700 on Build GRCh38
rs772706772 Chr.4: 186271713-186271713 on Build GRCh38
rs932824943 Chr.4: 186271642-186271642 on Build GRCh38
rs1002456131 Chr.4: 186271759-186271759 on Build GRCh38
rs121965069 Chr.4: 186271719-186271719 on Build GRCh38
rs775930271 Chr.4: 186271612-186271612 on Build GRCh38
rs751437478 Chr.4: 186271670-186271670 on Build GRCh38
rs771398102 Chr.4: 186271585-186271585 on Build GRCh38
rs55956266 Chr.4: 186271710-186271710 on Build GRCh38
rs768158918 Chr.4: 186271731-186271731 on Build GRCh38
rs1052041692 Chr.4: 186271685-186271685 on Build GRCh38
rs766389425 Chr.4: 186271762-186271762 on Build GRCh38
rs766464009 Chr.4: 186271586-186271586 on Build GRCh38
rs757279802 Chr.4: 186271691-186271691 on Build GRCh38
rs768409400 Chr.4: 186271620-186271620 on Build GRCh38
rs879034415 Chr.4: 186271629-186271629 on Build GRCh38
rs369667028 Chr.4: 186271779-186271779 on Build GRCh38
rs567451638 Chr.4: 186271768-186271768 on Build GRCh38
rs368700205 Chr.4: 186271793-186271793 on Build GRCh38
rs767240420 Chr.4: 186271775-186271775 on Build GRCh38
rs764869222 Chr.4: 186271651-186271651 on Build GRCh38
rs755319181 Chr.4: 186271794-186271794 on Build GRCh38
rs754973084 Chr.4: 186271679-186271679 on Build GRCh38
rs144595035 Chr.4: 186271749-186271749 on Build GRCh38
rs281875261 Chr.4: 186271712-186271712 on Build GRCh38
rs896571623 Chr.4: 186271770-186271770 on Build GRCh38
rs5968 Chr.4: 186271750-186271750 on Build GRCh38
rs211694396 Chr.4: 186271672-186271672 on Build GRCh38
rs773987040 Chr.4: 186271767-186271767 on Build GRCh38
rs281875271 Chr.4: 186271690-186271690 on Build GRCh38
rs11940999 Chr.4: 186271791-186271791 on Build GRCh38
rs281875264 Chr.4: 186271680-186271680 on Build GRCh38
rs779354401 Chr.4: 186271596-186271596 on Build GRCh38
rs368183250 Chr.4: 186271649-186271649 on Build GRCh38
rs372339062 Chr.4: 186271795-186271795 on Build GRCh38
rs281875267 Chr.4: 186271653-186271653 on Build GRCh38
rs747446747 Chr.4: 186271730-186271730 on Build GRCh38
rs374122923 Chr.4: 186271603-186271603 on Build GRCh38
rs533298073 Chr.4: 186271702-186271702 on Build GRCh38
rs772851488 Chr.4: 186271754-186271754 on Build GRCh38
rs746398473 Chr.4: 186271600-186271600 on Build GRCh38
rs140731806 Chr.4: 186271742-186271742 on Build GRCh38
rs780647948 Chr.4: 186271792-186271792 on Build GRCh38
rs1057516738 Chr.4: 186271772-186271772 on Build GRCh38
rs778981487 Chr.4: 186271694-186271694 on Build GRCh38
rs281875243 Chr.4: 186271704-186271704 on Build GRCh38
rs757477535 Chr.4: 186271782-186271782 on Build GRCh38
rs539655151 Chr.4: 186271683-186271683 on Build GRCh38
rs281875244 Chr.4: 186271741-186271741 on Build GRCh38
rs779497712 Chr.4: 186271769-186271769 on Build GRCh38
rs281875259 Chr.4: 186271647-186271647 on Build GRCh38

View Full Product Details