Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35266384-35266625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSR
Entrez Gene ID
51599
Gene Name
lipolysis stimulated lipoprotein receptor
Gene Aliases
ILDR3, LISCH7
UniGene
Hs.466507
Gene Chromosome Location
Chr.19: 35248656-35267964 on Build GRCh38
Transcripts
NM_001260489.1 NM_001260490.1 NM_015925.6 NM_205834.3 NM_205835.3 XM_005258980.1 XM_005258982.1 XM_011527026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
148590 c.1088G>A p.S363N Chr.19: 35266524-35266524 on Build GRCh38
3197509 c.1054T>C p.Y352H Chr.19: 35266490-35266490 on Build GRCh38
1712151 c.1026G>A p.M342I Chr.19: 35266462-35266462 on Build GRCh38
4580943 c.984T>C p.Y328Y Chr.19: 35266420-35266420 on Build GRCh38
3197507 c.1010C>A p.P337Q Chr.19: 35266446-35266446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001819832 Chr.19: 35266598-35266598 on Build GRCh38
rs201053883 Chr.19: 35266478-35266478 on Build GRCh38
rs765720719 Chr.19: 35266532-35266532 on Build GRCh38
rs768657158 Chr.19: 35266569-35266569 on Build GRCh38
rs745538204 Chr.19: 35266528-35266528 on Build GRCh38
rs760953725 Chr.19: 35266427-35266427 on Build GRCh38
rs373802174 Chr.19: 35266420-35266420 on Build GRCh38
rs766653513 Chr.19: 35266520-35266520 on Build GRCh38
rs780522833 Chr.19: 35266488-35266488 on Build GRCh38
rs776969335 Chr.19: 35266583-35266583 on Build GRCh38
rs770834401 Chr.19: 35266551-35266551 on Build GRCh38
rs112787787 Chr.19: 35266565-35266565 on Build GRCh38
rs556442317 Chr.19: 35266556-35266556 on Build GRCh38
rs766052565 Chr.19: 35266489-35266490 on Build GRCh38
rs764308558 Chr.19: 35266541-35266541 on Build GRCh38
rs769877684 Chr.19: 35266407-35266407 on Build GRCh38
rs774207346 Chr.19: 35266515-35266515 on Build GRCh38
rs370400208 Chr.19: 35266561-35266561 on Build GRCh38
rs368298403 Chr.19: 35266505-35266505 on Build GRCh38
rs201814864 Chr.19: 35266419-35266419 on Build GRCh38
rs766308951 Chr.19: 35266428-35266428 on Build GRCh38
rs773369862 Chr.19: 35266425-35266425 on Build GRCh38
rs912834366 Chr.19: 35266594-35266594 on Build GRCh38
rs34259399 Chr.19: 35266524-35266524 on Build GRCh38
rs758912303 Chr.19: 35266540-35266540 on Build GRCh38
rs536562931 Chr.19: 35266494-35266494 on Build GRCh38
rs773975196 Chr.19: 35266571-35266571 on Build GRCh38
rs745352489 Chr.19: 35266562-35266562 on Build GRCh38
rs148362066 Chr.19: 35266446-35266446 on Build GRCh38
rs768494910 Chr.19: 35266513-35266513 on Build GRCh38
rs905716218 Chr.19: 35266590-35266590 on Build GRCh38
rs758690987 Chr.19: 35266491-35266491 on Build GRCh38
rs188344681 Chr.19: 35266435-35266435 on Build GRCh38
rs769828448 Chr.19: 35266500-35266500 on Build GRCh38
rs759336923 Chr.19: 35266406-35266406 on Build GRCh38
rs753297763 Chr.19: 35266540-35266541 on Build GRCh38
rs879172684 Chr.19: 35266533-35266533 on Build GRCh38
rs1016484474 Chr.19: 35266586-35266586 on Build GRCh38
rs749086435 Chr.19: 35266506-35266506 on Build GRCh38
rs1035742967 Chr.19: 35266431-35266431 on Build GRCh38
rs755436260 Chr.19: 35266436-35266436 on Build GRCh38
rs760756509 Chr.19: 35266516-35266516 on Build GRCh38
rs142130344 Chr.19: 35266486-35266486 on Build GRCh38
rs12459658 Chr.19: 35266566-35266566 on Build GRCh38
rs756948996 Chr.19: 35266487-35266487 on Build GRCh38
rs751979420 Chr.19: 35266558-35266558 on Build GRCh38
rs575593310 Chr.19: 35266567-35266567 on Build GRCh38
rs1015591664 Chr.19: 35266525-35266525 on Build GRCh38
rs775389479 Chr.19: 35266416-35266416 on Build GRCh38
rs753038064 Chr.19: 35266448-35266448 on Build GRCh38
rs756823048 Chr.19: 35266560-35266560 on Build GRCh38
rs748012806 Chr.19: 35266581-35266581 on Build GRCh38
rs758822149 Chr.19: 35266463-35266463 on Build GRCh38
rs537487857 Chr.19: 35266461-35266461 on Build GRCh38
rs753269424 Chr.19: 35266537-35266537 on Build GRCh38

View Full Product Details