Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36758234-36758487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT27
Entrez Gene ID
11020
Gene Name
intraflagellar transport 27
Gene Aliases
BBS19, RABL4, RAYL
UniGene
Hs.415172
Gene Chromosome Location
Chr.22: 36758202-36776133 on Build GRCh38
Transcripts
NM_001177701.2 NM_006860.4 XM_006724106.2 XM_017028540.1 NR_134623.1

Gene Symbol
LOC105373021
Entrez Gene ID
105373021
Gene Name
uncharacterized LOC105373021
Gene Aliases
-
UniGene
Hs.401697
Gene Chromosome Location
Chr.22: 36703876-36767089 on Build GRCh38
Transcripts
NM_001177701.2 NM_006860.4 XM_006724106.2 XM_017028540.1 NR_134623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371831 c.538G>T p.V180F Chr.22: 36758331-36758331 on Build GRCh38
5916903 c.487T>A p.F163I Chr.22: 36758382-36758382 on Build GRCh38
1190437 c.481G>A p.A161T Chr.22: 36758388-36758388 on Build GRCh38
6317813 c.544C>A p.R182R Chr.22: 36758325-36758325 on Build GRCh38
3149869 c.545G>A p.R182Q Chr.22: 36758324-36758324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769556333 Chr.22: 36758453-36758453 on Build GRCh38
rs373124388 Chr.22: 36758281-36758281 on Build GRCh38
rs779134370 Chr.22: 36758407-36758407 on Build GRCh38
rs752867047 Chr.22: 36758408-36758408 on Build GRCh38
rs375934694 Chr.22: 36758280-36758280 on Build GRCh38
rs145066542 Chr.22: 36758362-36758362 on Build GRCh38
rs368923336 Chr.22: 36758310-36758310 on Build GRCh38
rs577272069 Chr.22: 36758424-36758424 on Build GRCh38
rs754968938 Chr.22: 36758318-36758318 on Build GRCh38
rs767854628 Chr.22: 36758297-36758297 on Build GRCh38
rs777805137 Chr.22: 36758422-36758422 on Build GRCh38
rs759231913 Chr.22: 36758357-36758357 on Build GRCh38
rs369096634 Chr.22: 36758374-36758374 on Build GRCh38
rs745493719 Chr.22: 36758439-36758439 on Build GRCh38
rs750851914 Chr.22: 36758305-36758305 on Build GRCh38
rs143144562 Chr.22: 36758392-36758392 on Build GRCh38
rs758390616 Chr.22: 36758411-36758411 on Build GRCh38
rs199622557 Chr.22: 36758452-36758452 on Build GRCh38
rs750761644 Chr.22: 36758375-36758375 on Build GRCh38
rs118095726 Chr.22: 36758369-36758369 on Build GRCh38
rs769127617 Chr.22: 36758359-36758359 on Build GRCh38
rs866263201 Chr.22: 36758316-36758316 on Build GRCh38
rs373367496 Chr.22: 36758460-36758460 on Build GRCh38
rs371608183 Chr.22: 36758436-36758436 on Build GRCh38
rs756296702 Chr.22: 36758309-36758309 on Build GRCh38
rs776108336 Chr.22: 36758354-36758354 on Build GRCh38
rs764835637 Chr.22: 36758294-36758294 on Build GRCh38
rs754110624 Chr.22: 36758314-36758314 on Build GRCh38
rs753938507 Chr.22: 36758391-36758391 on Build GRCh38
rs539071707 Chr.22: 36758378-36758378 on Build GRCh38
rs150387982 Chr.22: 36758325-36758325 on Build GRCh38
rs770564746 Chr.22: 36758430-36758430 on Build GRCh38
rs201002936 Chr.22: 36758429-36758429 on Build GRCh38
rs140440023 Chr.22: 36758324-36758324 on Build GRCh38
rs568701732 Chr.22: 36758345-36758345 on Build GRCh38
rs778064005 Chr.22: 36758293-36758293 on Build GRCh38
rs201534129 Chr.22: 36758282-36758282 on Build GRCh38
rs201189943 Chr.22: 36758346-36758346 on Build GRCh38
rs200349828 Chr.22: 36758295-36758295 on Build GRCh38
rs372265245 Chr.22: 36758275-36758275 on Build GRCh38
rs368963202 Chr.22: 36758273-36758273 on Build GRCh38
rs537237649 Chr.22: 36758350-36758350 on Build GRCh38
rs751795100 Chr.22: 36758305-36758306 on Build GRCh38
rs757405956 Chr.22: 36758436-36758437 on Build GRCh38
rs772206857 Chr.22: 36758337-36758337 on Build GRCh38
rs766794230 Chr.22: 36758380-36758380 on Build GRCh38
rs762301687 Chr.22: 36758367-36758367 on Build GRCh38

View Full Product Details