Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1353467-1353740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXRA8
Entrez Gene ID
54587
Gene Name
matrix remodeling associated 8
Gene Aliases
-
UniGene
Hs.515687
Gene Chromosome Location
Chr.1: 1352689-1363541 on Build GRCh38
Transcripts
NM_001282582.1 NM_001282583.1 NM_001282584.1 NM_001282585.1 NM_032348.3 XM_017001516.1 XM_017001517.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4525182 c.1314G>A p.K438K Chr.1: 1353619-1353619 on Build GRCh38
1333404 c.1315_1317delGAG p.E439delE Chr.1: 1353616-1353618 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769103152 Chr.1: 1353628-1353628 on Build GRCh38
rs775632584 Chr.1: 1353570-1353570 on Build GRCh38
rs773378933 Chr.1: 1353658-1353658 on Build GRCh38
rs762793523 Chr.1: 1353575-1353575 on Build GRCh38
rs572913095 Chr.1: 1353542-1353542 on Build GRCh38
rs977074787 Chr.1: 1353703-1353703 on Build GRCh38
rs201452853 Chr.1: 1353553-1353554 on Build GRCh38
rs757467726 Chr.1: 1353650-1353650 on Build GRCh38
rs979751953 Chr.1: 1353587-1353587 on Build GRCh38
rs552753693 Chr.1: 1353527-1353527 on Build GRCh38
rs766189200 Chr.1: 1353714-1353714 on Build GRCh38
rs1008064064 Chr.1: 1353675-1353675 on Build GRCh38
rs370804277 Chr.1: 1353622-1353622 on Build GRCh38
rs775860030 Chr.1: 1353583-1353583 on Build GRCh38
rs138720282 Chr.1: 1353493-1353493 on Build GRCh38
rs867566202 Chr.1: 1353610-1353610 on Build GRCh38
rs369865566 Chr.1: 1353707-1353708 on Build GRCh38
rs377351163 Chr.1: 1353649-1353649 on Build GRCh38
rs548757067 Chr.1: 1353556-1353557 on Build GRCh38
rs760167084 Chr.1: 1353673-1353673 on Build GRCh38
rs552424448 Chr.1: 1353694-1353694 on Build GRCh38
rs749797745 Chr.1: 1353624-1353624 on Build GRCh38
rs1026853060 Chr.1: 1353526-1353526 on Build GRCh38
rs866651627 Chr.1: 1353630-1353630 on Build GRCh38
rs768275416 Chr.1: 1353592-1353592 on Build GRCh38
rs1043529828 Chr.1: 1353618-1353618 on Build GRCh38
rs377381650 Chr.1: 1353613-1353613 on Build GRCh38
rs183058363 Chr.1: 1353645-1353645 on Build GRCh38
rs148936718 Chr.1: 1353546-1353546 on Build GRCh38
rs146120010 Chr.1: 1353663-1353663 on Build GRCh38
rs764911288 Chr.1: 1353616-1353618 on Build GRCh38
rs911114773 Chr.1: 1353547-1353547 on Build GRCh38
rs754362980 Chr.1: 1353626-1353626 on Build GRCh38
rs187396983 Chr.1: 1353657-1353657 on Build GRCh38
rs748024896 Chr.1: 1353661-1353661 on Build GRCh38
rs568314233 Chr.1: 1353496-1353496 on Build GRCh38
rs35421347 Chr.1: 1353550-1353550 on Build GRCh38
rs536172303 Chr.1: 1353516-1353516 on Build GRCh38
rs908004430 Chr.1: 1353614-1353614 on Build GRCh38
rs993042007 Chr.1: 1353717-1353717 on Build GRCh38
rs772916832 Chr.1: 1353672-1353672 on Build GRCh38
rs979889393 Chr.1: 1353707-1353707 on Build GRCh38
rs778177801 Chr.1: 1353646-1353646 on Build GRCh38
rs540440841 Chr.1: 1353651-1353651 on Build GRCh38
rs867755974 Chr.1: 1353636-1353636 on Build GRCh38
rs913267754 Chr.1: 1353705-1353705 on Build GRCh38
rs567316755 Chr.1: 1353700-1353701 on Build GRCh38
rs544512102 Chr.1: 1353631-1353631 on Build GRCh38
rs147014928 Chr.1: 1353603-1353603 on Build GRCh38
rs753141750 Chr.1: 1353644-1353644 on Build GRCh38
rs1056069518 Chr.1: 1353633-1353633 on Build GRCh38
rs965289438 Chr.1: 1353701-1353701 on Build GRCh38
rs534128898 Chr.1: 1353499-1353499 on Build GRCh38
rs1010499550 Chr.1: 1353701-1353701 on Build GRCh38
rs370687979 Chr.1: 1353659-1353659 on Build GRCh38
rs946194408 Chr.1: 1353606-1353606 on Build GRCh38
rs760218528 Chr.1: 1353623-1353623 on Build GRCh38
rs779084355 Chr.1: 1353642-1353642 on Build GRCh38
rs774199196 Chr.1: 1353595-1353595 on Build GRCh38
rs961177740 Chr.1: 1353505-1353505 on Build GRCh38
rs192450263 Chr.1: 1353557-1353557 on Build GRCh38
rs867158781 Chr.1: 1353561-1353561 on Build GRCh38
rs1019492290 Chr.1: 1353689-1353690 on Build GRCh38

View Full Product Details