Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177337616-177337857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMAN2
Entrez Gene ID
10960
Gene Name
lectin, mannose binding 2
Gene Aliases
C5orf8, GP36B, VIP36
UniGene
Hs.75864
Gene Chromosome Location
Chr.5: 177331562-177351884 on Build GRCh38
Transcripts
NM_006816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4898995 c.509C>T p.T170I Chr.5: 177337710-177337710 on Build GRCh38
4495881 c.462C>T p.F154F Chr.5: 177337757-177337757 on Build GRCh38
737586 c.435G>A p.G145G Chr.5: 177337784-177337784 on Build GRCh38
3614528 c.495C>T p.P165P Chr.5: 177337724-177337724 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1051385160 Chr.5: 177337796-177337796 on Build GRCh38
rs200117951 Chr.5: 177337831-177337831 on Build GRCh38
rs779183923 Chr.5: 177337774-177337774 on Build GRCh38
rs778971472 Chr.5: 177337691-177337691 on Build GRCh38
rs751488465 Chr.5: 177337731-177337731 on Build GRCh38
rs564394272 Chr.5: 177337655-177337655 on Build GRCh38
rs746002262 Chr.5: 177337783-177337783 on Build GRCh38
rs764922843 Chr.5: 177337721-177337721 on Build GRCh38
rs532080713 Chr.5: 177337656-177337656 on Build GRCh38
rs780478080 Chr.5: 177337805-177337805 on Build GRCh38
rs754768320 Chr.5: 177337680-177337680 on Build GRCh38
rs777089377 Chr.5: 177337713-177337713 on Build GRCh38
rs760985961 Chr.5: 177337711-177337711 on Build GRCh38
rs747577043 Chr.5: 177337830-177337830 on Build GRCh38
rs368405970 Chr.5: 177337815-177337815 on Build GRCh38
rs547485659 Chr.5: 177337798-177337798 on Build GRCh38
rs751376714 Chr.5: 177337679-177337679 on Build GRCh38
rs749959243 Chr.5: 177337722-177337722 on Build GRCh38
rs772517405 Chr.5: 177337701-177337701 on Build GRCh38
rs145658605 Chr.5: 177337738-177337738 on Build GRCh38
rs1047099183 Chr.5: 177337668-177337668 on Build GRCh38
rs780027669 Chr.5: 177337678-177337678 on Build GRCh38
rs200897982 Chr.5: 177337728-177337728 on Build GRCh38
rs146629866 Chr.5: 177337754-177337754 on Build GRCh38
rs745905437 Chr.5: 177337700-177337700 on Build GRCh38
rs779478834 Chr.5: 177337816-177337816 on Build GRCh38
rs771099552 Chr.5: 177337689-177337689 on Build GRCh38
rs780993962 Chr.5: 177337750-177337750 on Build GRCh38
rs769121880 Chr.5: 177337712-177337712 on Build GRCh38
rs371960732 Chr.5: 177337664-177337664 on Build GRCh38
rs543045024 Chr.5: 177337661-177337661 on Build GRCh38
rs762519854 Chr.5: 177337727-177337727 on Build GRCh38
rs376320709 Chr.5: 177337767-177337767 on Build GRCh38
rs200683893 Chr.5: 177337684-177337684 on Build GRCh38
rs761560510 Chr.5: 177337717-177337717 on Build GRCh38
rs941505316 Chr.5: 177337687-177337687 on Build GRCh38
rs1040264839 Chr.5: 177337692-177337692 on Build GRCh38
rs749532100 Chr.5: 177337688-177337688 on Build GRCh38

View Full Product Details