Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36498139-36498412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO19
Entrez Gene ID
80179
Gene Name
myosin XIX
Gene Aliases
MYOHD1
UniGene
Hs.302051
Gene Chromosome Location
Chr.17: 36495636-36535457 on Build GRCh38
Transcripts
NM_001163735.1 NM_025109.5 XM_011525286.1 XM_011525287.1 XM_011525290.2 XM_017025157.1 XM_017025158.1 XM_017025159.1 XM_017025160.1 XM_017025161.1 NM_001281432.1 NR_104011.1

Gene Symbol
ZNHIT3
Entrez Gene ID
9326
Gene Name
zinc finger HIT-type containing 3
Gene Aliases
TRIP3
UniGene
Hs.2210
Gene Chromosome Location
Chr.17: 36486627-36499310 on Build GRCh38
Transcripts
NM_001163735.1 NM_025109.5 XM_011525286.1 XM_011525287.1 XM_011525290.2 XM_017025157.1 XM_017025158.1 XM_017025159.1 XM_017025160.1 XM_017025161.1 NM_001281432.1 NR_104011.1

SNP Detail

SNP ID Chromosome Location
rs759830813 Chr.17: 36498223-36498223 on Build GRCh38
rs891961525 Chr.17: 36498390-36498390 on Build GRCh38
rs376647825 Chr.17: 36498256-36498256 on Build GRCh38
rs750771611 Chr.17: 36498318-36498318 on Build GRCh38
rs754700889 Chr.17: 36498257-36498257 on Build GRCh38
rs775493981 Chr.17: 36498355-36498355 on Build GRCh38
rs374149030 Chr.17: 36498205-36498205 on Build GRCh38
rs781533789 Chr.17: 36498344-36498344 on Build GRCh38
rs758074966 Chr.17: 36498314-36498314 on Build GRCh38
rs371087808 Chr.17: 36498181-36498181 on Build GRCh38
rs763351413 Chr.17: 36498294-36498297 on Build GRCh38
rs377461245 Chr.17: 36498261-36498261 on Build GRCh38
rs778418506 Chr.17: 36498307-36498307 on Build GRCh38
rs750368335 Chr.17: 36498247-36498247 on Build GRCh38
rs554560032 Chr.17: 36498294-36498294 on Build GRCh38
rs766196541 Chr.17: 36498288-36498288 on Build GRCh38
rs553297506 Chr.17: 36498250-36498250 on Build GRCh38
rs752409410 Chr.17: 36498312-36498312 on Build GRCh38
rs771513605 Chr.17: 36498343-36498343 on Build GRCh38
rs141763632 Chr.17: 36498262-36498262 on Build GRCh38
rs889893403 Chr.17: 36498367-36498367 on Build GRCh38
rs553814007 Chr.17: 36498254-36498254 on Build GRCh38
rs568029123 Chr.17: 36498239-36498239 on Build GRCh38
rs1019765902 Chr.17: 36498219-36498219 on Build GRCh38
rs778224764 Chr.17: 36498265-36498265 on Build GRCh38
rs1049840157 Chr.17: 36498170-36498170 on Build GRCh38
rs139565052 Chr.17: 36498277-36498277 on Build GRCh38
rs758408028 Chr.17: 36498280-36498280 on Build GRCh38
rs763188655 Chr.17: 36498365-36498365 on Build GRCh38
rs377403029 Chr.17: 36498284-36498284 on Build GRCh38
rs1035703594 Chr.17: 36498270-36498270 on Build GRCh38
rs1008823477 Chr.17: 36498194-36498194 on Build GRCh38
rs759508601 Chr.17: 36498275-36498275 on Build GRCh38
rs778312949 Chr.17: 36498334-36498334 on Build GRCh38
rs754434538 Chr.17: 36498298-36498298 on Build GRCh38
rs1041644283 Chr.17: 36498387-36498387 on Build GRCh38
rs768350186 Chr.17: 36498280-36498283 on Build GRCh38
rs117037794 Chr.17: 36498245-36498245 on Build GRCh38
rs934508641 Chr.17: 36498182-36498182 on Build GRCh38
rs768830269 Chr.17: 36498369-36498369 on Build GRCh38
rs962453818 Chr.17: 36498347-36498347 on Build GRCh38
rs773031246 Chr.17: 36498384-36498384 on Build GRCh38
rs760637363 Chr.17: 36498285-36498285 on Build GRCh38
rs370506869 Chr.17: 36498353-36498353 on Build GRCh38
rs867184489 Chr.17: 36498372-36498372 on Build GRCh38
rs745335102 Chr.17: 36498279-36498279 on Build GRCh38
rs574775346 Chr.17: 36498309-36498309 on Build GRCh38
rs538391929 Chr.17: 36498391-36498391 on Build GRCh38
rs765342710 Chr.17: 36498231-36498231 on Build GRCh38
rs968363092 Chr.17: 36498267-36498267 on Build GRCh38
rs773488388 Chr.17: 36498288-36498289 on Build GRCh38
rs1052502373 Chr.17: 36498183-36498183 on Build GRCh38
rs531394270 Chr.17: 36498171-36498171 on Build GRCh38
rs150601453 Chr.17: 36498268-36498268 on Build GRCh38
rs544274769 Chr.17: 36498283-36498283 on Build GRCh38
rs751416934 Chr.17: 36498233-36498233 on Build GRCh38
rs746023508 Chr.17: 36498345-36498345 on Build GRCh38
rs747539948 Chr.17: 36498266-36498266 on Build GRCh38
rs747341790 Chr.17: 36498336-36498336 on Build GRCh38

View Full Product Details