Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99885487-99885736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMTOR3
Entrez Gene ID
8649
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Gene Aliases
MAP2K1IP1, MAPBP, MAPKSP1, MP1, PRO0633, Ragulator3
UniGene
Hs.433332
Gene Chromosome Location
Chr.4: 99878338-99894546 on Build GRCh38
Transcripts
NM_001243736.1 NM_021970.3 NR_024170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1426092 c.136C>T p.R46* Chr.4: 99885643-99885643 on Build GRCh38
6269259 c.235C>A p.Q79K Chr.4: 99885544-99885544 on Build GRCh38
1049470 c.167C>T p.A56V Chr.4: 99885612-99885612 on Build GRCh38
4469946 c.162C>T p.A54A Chr.4: 99885617-99885617 on Build GRCh38
6582895 c.211A>G p.I71V Chr.4: 99885568-99885568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772205979 Chr.4: 99885551-99885551 on Build GRCh38
rs756416842 Chr.4: 99885637-99885637 on Build GRCh38
rs535019811 Chr.4: 99885688-99885688 on Build GRCh38
rs17852372 Chr.4: 99885644-99885644 on Build GRCh38
rs764756068 Chr.4: 99885557-99885557 on Build GRCh38
rs140420558 Chr.4: 99885663-99885663 on Build GRCh38
rs561188173 Chr.4: 99885555-99885555 on Build GRCh38
rs759206581 Chr.4: 99885709-99885709 on Build GRCh38
rs11546701 Chr.4: 99885639-99885639 on Build GRCh38
rs755204406 Chr.4: 99885649-99885649 on Build GRCh38
rs750862405 Chr.4: 99885628-99885628 on Build GRCh38
rs974173486 Chr.4: 99885651-99885651 on Build GRCh38
rs772114549 Chr.4: 99885661-99885661 on Build GRCh38
rs200997202 Chr.4: 99885571-99885571 on Build GRCh38
rs770680589 Chr.4: 99885681-99885681 on Build GRCh38
rs113542593 Chr.4: 99885520-99885520 on Build GRCh38
rs183234371 Chr.4: 99885566-99885566 on Build GRCh38
rs746869924 Chr.4: 99885677-99885677 on Build GRCh38
rs780467121 Chr.4: 99885642-99885642 on Build GRCh38
rs573265787 Chr.4: 99885647-99885647 on Build GRCh38
rs865943605 Chr.4: 99885618-99885618 on Build GRCh38
rs962384800 Chr.4: 99885643-99885643 on Build GRCh38
rs767841855 Chr.4: 99885619-99885619 on Build GRCh38
rs745903196 Chr.4: 99885515-99885515 on Build GRCh38
rs147607729 Chr.4: 99885658-99885658 on Build GRCh38
rs770675182 Chr.4: 99885525-99885525 on Build GRCh38
rs977856763 Chr.4: 99885563-99885563 on Build GRCh38
rs369101249 Chr.4: 99885526-99885526 on Build GRCh38
rs779148848 Chr.4: 99885652-99885652 on Build GRCh38
rs199578603 Chr.4: 99885573-99885573 on Build GRCh38
rs990194450 Chr.4: 99885513-99885513 on Build GRCh38
rs868690425 Chr.4: 99885582-99885582 on Build GRCh38
rs762392940 Chr.4: 99885568-99885568 on Build GRCh38

View Full Product Details