Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30803926-30804181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF3
Entrez Gene ID
51379
Gene Name
cytokine receptor like factor 3
Gene Aliases
CREME-9, CREME9, CRLM9, CYTOR4, FRWS, p48.2
UniGene
Hs.649372
Gene Chromosome Location
Chr.17: 30782684-30824760 on Build GRCh38
Transcripts
NM_015986.3 NR_073118.1 XM_011524879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3179332 c.303C>T p.H101H Chr.17: 30803935-30803935 on Build GRCh38
1630119 c.225G>A p.V75V Chr.17: 30804013-30804013 on Build GRCh38
7194830 c.220T>C p.L74L Chr.17: 30804018-30804018 on Build GRCh38
3179334 c.165C>G p.L55L Chr.17: 30804073-30804073 on Build GRCh38
977331 c.217C>T p.R73* Chr.17: 30804021-30804021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756235929 Chr.17: 30804078-30804078 on Build GRCh38
rs779345733 Chr.17: 30804116-30804116 on Build GRCh38
rs1031076018 Chr.17: 30804091-30804091 on Build GRCh38
rs775384003 Chr.17: 30804023-30804023 on Build GRCh38
rs751247850 Chr.17: 30804041-30804041 on Build GRCh38
rs767842783 Chr.17: 30804043-30804043 on Build GRCh38
rs752476295 Chr.17: 30804059-30804059 on Build GRCh38
rs770510980 Chr.17: 30803964-30803964 on Build GRCh38
rs144562177 Chr.17: 30803979-30803979 on Build GRCh38
rs763982011 Chr.17: 30804038-30804038 on Build GRCh38
rs776174924 Chr.17: 30804015-30804015 on Build GRCh38
rs754396895 Chr.17: 30804083-30804083 on Build GRCh38
rs769762917 Chr.17: 30804020-30804020 on Build GRCh38
rs769084122 Chr.17: 30803951-30803951 on Build GRCh38
rs999560735 Chr.17: 30804059-30804059 on Build GRCh38
rs112220518 Chr.17: 30803976-30803976 on Build GRCh38
rs780791493 Chr.17: 30804138-30804138 on Build GRCh38
rs117171945 Chr.17: 30804047-30804047 on Build GRCh38
rs996061068 Chr.17: 30804005-30804005 on Build GRCh38
rs35048389 Chr.17: 30804049-30804050 on Build GRCh38
rs774536161 Chr.17: 30803956-30803956 on Build GRCh38
rs377477807 Chr.17: 30804025-30804025 on Build GRCh38
rs762111144 Chr.17: 30804042-30804042 on Build GRCh38
rs755371282 Chr.17: 30804098-30804098 on Build GRCh38
rs773415390 Chr.17: 30804079-30804079 on Build GRCh38
rs748446679 Chr.17: 30804117-30804117 on Build GRCh38
rs760653755 Chr.17: 30804121-30804125 on Build GRCh38
rs770635588 Chr.17: 30804135-30804135 on Build GRCh38
rs780229090 Chr.17: 30804080-30804080 on Build GRCh38
rs1002751844 Chr.17: 30804030-30804030 on Build GRCh38
rs527256643 Chr.17: 30804103-30804103 on Build GRCh38
rs764061404 Chr.17: 30804148-30804148 on Build GRCh38
rs759042791 Chr.17: 30804018-30804018 on Build GRCh38
rs954710361 Chr.17: 30804028-30804028 on Build GRCh38
rs745430664 Chr.17: 30804141-30804141 on Build GRCh38
rs370679974 Chr.17: 30804081-30804081 on Build GRCh38

View Full Product Details