Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94562468-94562729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELLS
Entrez Gene ID
3070
Gene Name
helicase, lymphoid-specific
Gene Aliases
ICF4, LSH, Nbla10143, PASG, SMARCA6
UniGene
Hs.463677
Gene Chromosome Location
Chr.10: 94545767-94613905 on Build GRCh38
Transcripts
NM_001289067.1 NM_001289068.1 NM_001289069.1 NM_001289070.1 NM_001289071.1 NM_001289072.1 NM_001289073.1 NM_001289074.1 NM_001289075.1 NM_018063.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209512 c.347T>C p.I116T Chr.10: 94562704-94562704 on Build GRCh38
7370945 c.334G>A p.G112S Chr.10: 94562691-94562691 on Build GRCh38
921713 c.340A>C p.N114H Chr.10: 94562697-94562697 on Build GRCh38
7370946 c.334G>A p.G112S Chr.10: 94562691-94562691 on Build GRCh38
5718950 c.344C>T p.S115L Chr.10: 94562701-94562701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200134892 Chr.10: 94562704-94562704 on Build GRCh38
rs188955699 Chr.10: 94562672-94562672 on Build GRCh38
rs141686502 Chr.10: 94562700-94562700 on Build GRCh38
rs888754330 Chr.10: 94562707-94562707 on Build GRCh38
rs17110134 Chr.10: 94562556-94562556 on Build GRCh38
rs948852469 Chr.10: 94562601-94562602 on Build GRCh38
rs759447822 Chr.10: 94562642-94562642 on Build GRCh38
rs10882477 Chr.10: 94562647-94562647 on Build GRCh38
rs184503814 Chr.10: 94562609-94562609 on Build GRCh38
rs148139900 Chr.10: 94562691-94562691 on Build GRCh38
rs752822423 Chr.10: 94562670-94562670 on Build GRCh38
rs934850772 Chr.10: 94562637-94562637 on Build GRCh38
rs1051318033 Chr.10: 94562639-94562639 on Build GRCh38
rs971856494 Chr.10: 94562519-94562519 on Build GRCh38
rs200659324 Chr.10: 94562698-94562698 on Build GRCh38
rs764213137 Chr.10: 94562678-94562678 on Build GRCh38
rs939393446 Chr.10: 94562551-94562551 on Build GRCh38
rs781722925 Chr.10: 94562686-94562686 on Build GRCh38
rs778345468 Chr.10: 94562597-94562597 on Build GRCh38
rs147153698 Chr.10: 94562703-94562703 on Build GRCh38
rs868009906 Chr.10: 94562587-94562587 on Build GRCh38
rs753956049 Chr.10: 94562685-94562685 on Build GRCh38
rs1007122018 Chr.10: 94562663-94562663 on Build GRCh38
rs559271075 Chr.10: 94562635-94562635 on Build GRCh38
rs994289499 Chr.10: 94562623-94562623 on Build GRCh38
rs759796016 Chr.10: 94562658-94562658 on Build GRCh38
rs1038382456 Chr.10: 94562601-94562601 on Build GRCh38
rs916561966 Chr.10: 94562589-94562589 on Build GRCh38
rs966294784 Chr.10: 94562501-94562501 on Build GRCh38
rs561336502 Chr.10: 94562693-94562693 on Build GRCh38
rs767945866 Chr.10: 94562659-94562659 on Build GRCh38
rs928526922 Chr.10: 94562546-94562546 on Build GRCh38
rs557239854 Chr.10: 94562491-94562491 on Build GRCh38

View Full Product Details