Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111217847-111218080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUX2
Entrez Gene ID
23316
Gene Name
cut like homeobox 2
Gene Aliases
CDP2, CUTL2
UniGene
Hs.124953
Gene Chromosome Location
Chr.12: 111034024-111350556 on Build GRCh38
Transcripts
NM_015267.3 XM_011538061.2 XM_011538063.2 XM_011538069.2 XM_011538070.2 XM_017019080.1 XM_017019081.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5374162 c.218_219CC>TT p.A73V Chr.12: 111217933-111217934 on Build GRCh38
5920398 c.214C>T p.Q72* Chr.12: 111217929-111217929 on Build GRCh38
415625 c.222+1G>T p.? Chr.12: 111217938-111217938 on Build GRCh38
4929098 c.215A>T p.Q72L Chr.12: 111217930-111217930 on Build GRCh38
5724145 c.186G>A p.E62E Chr.12: 111217901-111217901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532299907 Chr.12: 111218016-111218016 on Build GRCh38
rs749500899 Chr.12: 111217957-111217957 on Build GRCh38
rs776310745 Chr.12: 111217978-111217978 on Build GRCh38
rs1013110630 Chr.12: 111217881-111217881 on Build GRCh38
rs564823732 Chr.12: 111217992-111217992 on Build GRCh38
rs78545855 Chr.12: 111217948-111217948 on Build GRCh38
rs750419179 Chr.12: 111217876-111217876 on Build GRCh38
rs766887760 Chr.12: 111217950-111217950 on Build GRCh38
rs745896959 Chr.12: 111217961-111217961 on Build GRCh38
rs762466529 Chr.12: 111217989-111217989 on Build GRCh38
rs1033269679 Chr.12: 111217965-111217965 on Build GRCh38
rs761440308 Chr.12: 111217979-111217979 on Build GRCh38
rs751335439 Chr.12: 111217935-111217935 on Build GRCh38
rs915097305 Chr.12: 111218021-111218021 on Build GRCh38
rs147588505 Chr.12: 111217960-111217960 on Build GRCh38
rs766271204 Chr.12: 111217931-111217931 on Build GRCh38
rs867450452 Chr.12: 111217997-111217997 on Build GRCh38
rs547308565 Chr.12: 111218019-111218019 on Build GRCh38
rs776013717 Chr.12: 111217972-111217972 on Build GRCh38
rs992269535 Chr.12: 111217985-111217985 on Build GRCh38
rs61073405 Chr.12: 111218044-111218044 on Build GRCh38
rs764793527 Chr.12: 111217980-111217980 on Build GRCh38
rs200060130 Chr.12: 111217974-111217974 on Build GRCh38
rs772711257 Chr.12: 111217988-111217988 on Build GRCh38
rs576157014 Chr.12: 111217963-111217963 on Build GRCh38
rs989217151 Chr.12: 111218003-111218003 on Build GRCh38
rs1004031541 Chr.12: 111217896-111217896 on Build GRCh38
rs758261866 Chr.12: 111217928-111217928 on Build GRCh38
rs754764523 Chr.12: 111217944-111217944 on Build GRCh38

View Full Product Details