Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77025356-77025593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF2BPL
Entrez Gene ID
64207
Gene Name
interferon regulatory factor 2 binding protein like
Gene Aliases
C14orf4, EAP1
UniGene
Hs.179260
Gene Chromosome Location
Chr.14: 77024543-77028699 on Build GRCh38
Transcripts
NM_024496.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607989 c.2332G>T p.G778C Chr.14: 77025461-77025461 on Build GRCh38
1562526 c.2334C>T p.G778G Chr.14: 77025459-77025459 on Build GRCh38
5493206 c.2280C>T p.S760S Chr.14: 77025513-77025513 on Build GRCh38
6687242 c.2255C>G p.A752G Chr.14: 77025538-77025538 on Build GRCh38
6035095 c.2307G>T p.S769S Chr.14: 77025486-77025486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760939648 Chr.14: 77025492-77025492 on Build GRCh38
rs765646476 Chr.14: 77025387-77025387 on Build GRCh38
rs894267486 Chr.14: 77025504-77025504 on Build GRCh38
rs151294479 Chr.14: 77025516-77025516 on Build GRCh38
rs903060962 Chr.14: 77025453-77025453 on Build GRCh38
rs765342094 Chr.14: 77025399-77025400 on Build GRCh38
rs771978372 Chr.14: 77025567-77025567 on Build GRCh38
rs748033590 Chr.14: 77025558-77025558 on Build GRCh38
rs1020170411 Chr.14: 77025438-77025438 on Build GRCh38
rs753122396 Chr.14: 77025405-77025405 on Build GRCh38
rs201025100 Chr.14: 77025446-77025446 on Build GRCh38
rs945508002 Chr.14: 77025407-77025407 on Build GRCh38
rs199906186 Chr.14: 77025382-77025382 on Build GRCh38
rs778657493 Chr.14: 77025471-77025471 on Build GRCh38
rs760847179 Chr.14: 77025485-77025485 on Build GRCh38
rs142468978 Chr.14: 77025494-77025494 on Build GRCh38
rs368055469 Chr.14: 77025535-77025535 on Build GRCh38
rs577997949 Chr.14: 77025531-77025531 on Build GRCh38
rs189074743 Chr.14: 77025451-77025451 on Build GRCh38
rs776826063 Chr.14: 77025495-77025495 on Build GRCh38
rs201849597 Chr.14: 77025383-77025383 on Build GRCh38
rs145752129 Chr.14: 77025486-77025486 on Build GRCh38
rs759473791 Chr.14: 77025499-77025499 on Build GRCh38
rs758765386 Chr.14: 77025408-77025408 on Build GRCh38
rs764601763 Chr.14: 77025520-77025520 on Build GRCh38
rs779653559 Chr.14: 77025450-77025450 on Build GRCh38
rs914060717 Chr.14: 77025388-77025388 on Build GRCh38
rs778857044 Chr.14: 77025547-77025547 on Build GRCh38
rs765774796 Chr.14: 77025514-77025514 on Build GRCh38
rs745468671 Chr.14: 77025444-77025444 on Build GRCh38
rs751939199 Chr.14: 77025525-77025525 on Build GRCh38
rs935659940 Chr.14: 77025497-77025497 on Build GRCh38
rs149689849 Chr.14: 77025546-77025546 on Build GRCh38
rs768698108 Chr.14: 77025452-77025452 on Build GRCh38
rs781023065 Chr.14: 77025417-77025417 on Build GRCh38
rs1055459865 Chr.14: 77025500-77025500 on Build GRCh38
rs115356554 Chr.14: 77025519-77025519 on Build GRCh38

View Full Product Details