Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38538212-38538461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMC1
Entrez Gene ID
11144
Gene Name
DNA meiotic recombinase 1
Gene Aliases
DMC1H, LIM15, dJ199H16.1
UniGene
Hs.339396
Gene Chromosome Location
Chr.22: 38509154-38570286 on Build GRCh38
Transcripts
NM_001278208.1 NM_007068.3 XM_006724112.2 XM_011529834.1 XM_011529835.2 XM_011529837.1 XM_011529838.1 XM_017028542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034153 c.740C>T p.A247V Chr.22: 38538330-38538330 on Build GRCh38
4977707 c.666C>T p.I222I Chr.22: 38538404-38538404 on Build GRCh38
3912799 c.703G>T p.G235C Chr.22: 38538367-38538367 on Build GRCh38
7140302 c.667G>T p.D223Y Chr.22: 38538403-38538403 on Build GRCh38
6468161 c.734A>T p.K245I Chr.22: 38538336-38538336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201179906 Chr.22: 38538288-38538288 on Build GRCh38
rs762406860 Chr.22: 38538303-38538304 on Build GRCh38
rs773347078 Chr.22: 38538418-38538418 on Build GRCh38
rs200187131 Chr.22: 38538294-38538294 on Build GRCh38
rs368393201 Chr.22: 38538248-38538248 on Build GRCh38
rs767965966 Chr.22: 38538372-38538372 on Build GRCh38
rs1001407956 Chr.22: 38538426-38538426 on Build GRCh38
rs760981205 Chr.22: 38538417-38538417 on Build GRCh38
rs751890214 Chr.22: 38538267-38538267 on Build GRCh38
rs766701089 Chr.22: 38538258-38538258 on Build GRCh38
rs761687068 Chr.22: 38538250-38538250 on Build GRCh38
rs138352828 Chr.22: 38538350-38538350 on Build GRCh38
rs201348870 Chr.22: 38538404-38538404 on Build GRCh38
rs765308228 Chr.22: 38538251-38538251 on Build GRCh38
rs867267644 Chr.22: 38538355-38538355 on Build GRCh38
rs767781951 Chr.22: 38538274-38538274 on Build GRCh38
rs755315236 Chr.22: 38538271-38538271 on Build GRCh38
rs746644230 Chr.22: 38538379-38538379 on Build GRCh38
rs757972386 Chr.22: 38538349-38538349 on Build GRCh38
rs143874492 Chr.22: 38538397-38538397 on Build GRCh38
rs199821104 Chr.22: 38538391-38538391 on Build GRCh38
rs1000731986 Chr.22: 38538362-38538362 on Build GRCh38
rs756601168 Chr.22: 38538308-38538308 on Build GRCh38
rs748020628 Chr.22: 38538403-38538403 on Build GRCh38
rs763113866 Chr.22: 38538424-38538424 on Build GRCh38
rs763162657 Chr.22: 38538255-38538255 on Build GRCh38
rs778302919 Chr.22: 38538315-38538315 on Build GRCh38
rs557897016 Chr.22: 38538253-38538253 on Build GRCh38
rs766582513 Chr.22: 38538425-38538425 on Build GRCh38
rs773429160 Chr.22: 38538416-38538416 on Build GRCh38
rs199712228 Chr.22: 38538329-38538329 on Build GRCh38

View Full Product Details