Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129176770-129176997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IER5L
Entrez Gene ID
389792
Gene Name
immediate early response 5 like
Gene Aliases
bA247A12.2
UniGene
Hs.529857
Gene Chromosome Location
Chr.9: 129175552-129178261 on Build GRCh38
Transcripts
NM_203434.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639266 c.1148G>A p.G383E Chr.9: 129176905-129176905 on Build GRCh38
3699463 c.1206C>G p.V402V Chr.9: 129176847-129176847 on Build GRCh38
5966499 c.1057A>T p.I353F Chr.9: 129176996-129176996 on Build GRCh38
4138850 c.1086C>G p.S362S Chr.9: 129176967-129176967 on Build GRCh38
5966500 c.702A>T p.T234T Chr.9: 129176996-129176996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776936905 Chr.9: 129176920-129176920 on Build GRCh38
rs752837307 Chr.9: 129176826-129176826 on Build GRCh38
rs759081248 Chr.9: 129176834-129176834 on Build GRCh38
rs2274722 Chr.9: 129176847-129176847 on Build GRCh38
rs559068583 Chr.9: 129176796-129176796 on Build GRCh38
rs764444653 Chr.9: 129176921-129176921 on Build GRCh38
rs954107862 Chr.9: 129176801-129176801 on Build GRCh38
rs778336057 Chr.9: 129176853-129176853 on Build GRCh38
rs929980424 Chr.9: 129176949-129176949 on Build GRCh38
rs754537722 Chr.9: 129176852-129176852 on Build GRCh38
rs750017430 Chr.9: 129176814-129176831 on Build GRCh38
rs779794793 Chr.9: 129176838-129176838 on Build GRCh38
rs758909167 Chr.9: 129176829-129176829 on Build GRCh38
rs760241978 Chr.9: 129176875-129176876 on Build GRCh38
rs762268207 Chr.9: 129176945-129176945 on Build GRCh38
rs762970820 Chr.9: 129176964-129176964 on Build GRCh38
rs751980672 Chr.9: 129176938-129176938 on Build GRCh38
rs938805607 Chr.9: 129176968-129176968 on Build GRCh38
rs757878512 Chr.9: 129176967-129176967 on Build GRCh38
rs568284969 Chr.9: 129176970-129176970 on Build GRCh38
rs926284281 Chr.9: 129176943-129176943 on Build GRCh38
rs200642588 Chr.9: 129176837-129176837 on Build GRCh38
rs746867931 Chr.9: 129176895-129176895 on Build GRCh38
rs752030835 Chr.9: 129176836-129176836 on Build GRCh38
rs987278902 Chr.9: 129176815-129176815 on Build GRCh38
rs772060335 Chr.9: 129176883-129176883 on Build GRCh38
rs759695858 Chr.9: 129176816-129176816 on Build GRCh38
rs1049003806 Chr.9: 129176856-129176856 on Build GRCh38
rs772920894 Chr.9: 129176790-129176790 on Build GRCh38
rs770595002 Chr.9: 129176908-129176908 on Build GRCh38
rs759623008 Chr.9: 129176922-129176922 on Build GRCh38
rs201443650 Chr.9: 129176795-129176795 on Build GRCh38
rs748078472 Chr.9: 129176871-129176871 on Build GRCh38
rs767262011 Chr.9: 129176813-129176830 on Build GRCh38
rs753540682 Chr.9: 129176955-129176955 on Build GRCh38
rs753273758 Chr.9: 129176824-129176824 on Build GRCh38
rs767723096 Chr.9: 129176952-129176952 on Build GRCh38
rs764748740 Chr.9: 129176935-129176935 on Build GRCh38
rs764693733 Chr.9: 129176831-129176831 on Build GRCh38
rs770002543 Chr.9: 129176931-129176931 on Build GRCh38
rs754558380 Chr.9: 129176961-129176961 on Build GRCh38
rs775779772 Chr.9: 129176932-129176932 on Build GRCh38
rs752259268 Chr.9: 129176966-129176966 on Build GRCh38
rs776692074 Chr.9: 129176797-129176797 on Build GRCh38
rs763113414 Chr.9: 129176933-129176933 on Build GRCh38

View Full Product Details