Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109608570-109608843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT2
Entrez Gene ID
2780
Gene Name
G protein subunit alpha transducin 2
Gene Aliases
ACHM4, GNATC
UniGene
Hs.36973
Gene Chromosome Location
Chr.1: 109603091-109619733 on Build GRCh38
Transcripts
NM_005272.3 XM_011541264.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3721940 c.309C>T p.D103D Chr.1: 109608783-109608783 on Build GRCh38
462554 c.308A>G p.D103G Chr.1: 109608784-109608784 on Build GRCh38
893546 c.314G>A p.R105Q Chr.1: 109608778-109608778 on Build GRCh38
1688232 c.350G>A p.G117E Chr.1: 109608742-109608742 on Build GRCh38
1636673 c.461+5G>C p.? Chr.1: 109608626-109608626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770564093 Chr.1: 109608628-109608628 on Build GRCh38
rs1029294333 Chr.1: 109608636-109608636 on Build GRCh38
rs754150816 Chr.1: 109608695-109608695 on Build GRCh38
rs973809932 Chr.1: 109608686-109608686 on Build GRCh38
rs772065303 Chr.1: 109608604-109608604 on Build GRCh38
rs747991149 Chr.1: 109608599-109608599 on Build GRCh38
rs976062103 Chr.1: 109608618-109608618 on Build GRCh38
rs773830399 Chr.1: 109608765-109608765 on Build GRCh38
rs760803840 Chr.1: 109608769-109608769 on Build GRCh38
rs752430152 Chr.1: 109608783-109608783 on Build GRCh38
rs755278059 Chr.1: 109608696-109608696 on Build GRCh38
rs762247888 Chr.1: 109608759-109608759 on Build GRCh38
rs753984783 Chr.1: 109608776-109608776 on Build GRCh38
rs758270188 Chr.1: 109608798-109608798 on Build GRCh38
rs1043276813 Chr.1: 109608763-109608763 on Build GRCh38
rs536959939 Chr.1: 109608672-109608672 on Build GRCh38
rs756332145 Chr.1: 109608680-109608680 on Build GRCh38
rs752549211 Chr.1: 109608706-109608706 on Build GRCh38
rs775374577 Chr.1: 109608751-109608751 on Build GRCh38
rs775416035 Chr.1: 109608815-109608817 on Build GRCh38
rs41280330 Chr.1: 109608722-109608722 on Build GRCh38
rs772456624 Chr.1: 109608762-109608762 on Build GRCh38
rs746532855 Chr.1: 109608622-109608622 on Build GRCh38
rs142976178 Chr.1: 109608750-109608750 on Build GRCh38
rs763283639 Chr.1: 109608642-109608642 on Build GRCh38
rs966097469 Chr.1: 109608606-109608606 on Build GRCh38
rs1054577802 Chr.1: 109608808-109608808 on Build GRCh38
rs777548321 Chr.1: 109608800-109608800 on Build GRCh38
rs764484604 Chr.1: 109608653-109608653 on Build GRCh38
rs3738766 Chr.1: 109608773-109608773 on Build GRCh38
rs147269835 Chr.1: 109608743-109608743 on Build GRCh38
rs774809648 Chr.1: 109608658-109608658 on Build GRCh38
rs751557801 Chr.1: 109608810-109608810 on Build GRCh38
rs777831176 Chr.1: 109608717-109608717 on Build GRCh38
rs758202508 Chr.1: 109608708-109608708 on Build GRCh38
rs766682709 Chr.1: 109608688-109608688 on Build GRCh38
rs145002495 Chr.1: 109608782-109608782 on Build GRCh38
rs776322769 Chr.1: 109608638-109608638 on Build GRCh38
rs780793773 Chr.1: 109608739-109608739 on Build GRCh38
rs12046787 Chr.1: 109608723-109608723 on Build GRCh38
rs909311863 Chr.1: 109608669-109608669 on Build GRCh38
rs149421007 Chr.1: 109608665-109608665 on Build GRCh38
rs768086755 Chr.1: 109608671-109608671 on Build GRCh38
rs772538094 Chr.1: 109608621-109608621 on Build GRCh38
rs778799065 Chr.1: 109608705-109608705 on Build GRCh38
rs759770362 Chr.1: 109608778-109608778 on Build GRCh38
rs767424501 Chr.1: 109608723-109608724 on Build GRCh38
rs200489857 Chr.1: 109608818-109608818 on Build GRCh38
rs397515384 Chr.1: 109608607-109608607 on Build GRCh38

View Full Product Details