Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149884989-149885246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEA8
Entrez Gene ID
4107
Gene Name
MAGE family member A8
Gene Aliases
CT1.8, MAGE8
UniGene
Hs.522803
Gene Chromosome Location
Chr.X: 149881141-149885835 on Build GRCh38
Transcripts
NM_001166400.1 NM_001166401.1 NM_005364.4 XM_017029520.1 NR_102703.1

Gene Symbol
MAGEA8-AS1
Entrez Gene ID
101410537
Gene Name
MAGEA8 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.573643
Gene Chromosome Location
Chr.X: 149878763-149897006 on Build GRCh38
Transcripts
NM_001166400.1 NM_001166401.1 NM_005364.4 XM_017029520.1 NR_102703.1

SNP Detail

SNP ID Chromosome Location
rs782124820 Chr.X: 149885021-149885021 on Build GRCh38
rs782641116 Chr.X: 149885103-149885103 on Build GRCh38
rs368749900 Chr.X: 149885214-149885214 on Build GRCh38
rs143187496 Chr.X: 149885174-149885174 on Build GRCh38
rs149075416 Chr.X: 149885143-149885143 on Build GRCh38
rs45564732 Chr.X: 149885043-149885043 on Build GRCh38
rs45577435 Chr.X: 149885069-149885069 on Build GRCh38
rs45567349 Chr.X: 149885201-149885201 on Build GRCh38
rs781850959 Chr.X: 149885019-149885019 on Build GRCh38
rs45578436 Chr.X: 149885183-149885183 on Build GRCh38
rs374612442 Chr.X: 149885068-149885068 on Build GRCh38
rs1040224114 Chr.X: 149885094-149885094 on Build GRCh38
rs45453393 Chr.X: 149885030-149885030 on Build GRCh38
rs782812593 Chr.X: 149885014-149885014 on Build GRCh38
rs45500197 Chr.X: 149885028-149885028 on Build GRCh38
rs372766396 Chr.X: 149885061-149885061 on Build GRCh38
rs45497502 Chr.X: 149885088-149885088 on Build GRCh38
rs929885088 Chr.X: 149885056-149885056 on Build GRCh38
rs140453811 Chr.X: 149885190-149885190 on Build GRCh38
rs782177723 Chr.X: 149885162-149885162 on Build GRCh38
rs903158321 Chr.X: 149885151-149885151 on Build GRCh38
rs782111183 Chr.X: 149885204-149885204 on Build GRCh38
rs782035958 Chr.X: 149885175-149885175 on Build GRCh38
rs782116149 Chr.X: 149885182-149885182 on Build GRCh38
rs45557437 Chr.X: 149885156-149885156 on Build GRCh38
rs376840959 Chr.X: 149885064-149885064 on Build GRCh38
rs781942405 Chr.X: 149885083-149885083 on Build GRCh38
rs45578531 Chr.X: 149885180-149885180 on Build GRCh38
rs782737761 Chr.X: 149885144-149885144 on Build GRCh38
rs782525478 Chr.X: 149885029-149885029 on Build GRCh38
rs149643429 Chr.X: 149885121-149885121 on Build GRCh38
rs12010332 Chr.X: 149885189-149885189 on Build GRCh38
rs45479394 Chr.X: 149885040-149885040 on Build GRCh38
rs782773975 Chr.X: 149885087-149885087 on Build GRCh38
rs782143734 Chr.X: 149885220-149885220 on Build GRCh38

View Full Product Details