Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 41325495-41325768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA16
Entrez Gene ID
79612
Gene Name
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Gene Aliases
NARG1L
UniGene
Hs.512914
Gene Chromosome Location
Chr.13: 41311205-41377031 on Build GRCh38
Transcripts
NM_001110798.1 NM_018527.3 NM_024561.4 XM_006719866.2 XM_011535227.2 XM_011535228.1 XM_017020744.1 XM_017020745.1 XM_017020746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3813822 c.591T>C p.N197N Chr.13: 41325751-41325751 on Build GRCh38
3813820 c.591T>C p.N197N Chr.13: 41325751-41325751 on Build GRCh38
947454 c.597G>A p.V199V Chr.13: 41325757-41325757 on Build GRCh38
3813821 c.591T>C p.N197N Chr.13: 41325751-41325751 on Build GRCh38
947452 c.576G>T p.L192F Chr.13: 41325736-41325736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780671572 Chr.13: 41325745-41325745 on Build GRCh38
rs184284983 Chr.13: 41325584-41325584 on Build GRCh38
rs369727164 Chr.13: 41325698-41325698 on Build GRCh38
rs758675911 Chr.13: 41325657-41325657 on Build GRCh38
rs189437379 Chr.13: 41325730-41325730 on Build GRCh38
rs7358825 Chr.13: 41325618-41325618 on Build GRCh38
rs770133246 Chr.13: 41325686-41325686 on Build GRCh38
rs371327547 Chr.13: 41325666-41325666 on Build GRCh38
rs757616878 Chr.13: 41325667-41325667 on Build GRCh38
rs761147743 Chr.13: 41325687-41325687 on Build GRCh38
rs563328537 Chr.13: 41325740-41325740 on Build GRCh38
rs148219104 Chr.13: 41325687-41325687 on Build GRCh38
rs764624610 Chr.13: 41325738-41325740 on Build GRCh38
rs781709653 Chr.13: 41325672-41325672 on Build GRCh38
rs760213103 Chr.13: 41325539-41325539 on Build GRCh38
rs915960498 Chr.13: 41325581-41325581 on Build GRCh38
rs1049990519 Chr.13: 41325688-41325688 on Build GRCh38
rs752648999 Chr.13: 41325652-41325652 on Build GRCh38
rs1039613281 Chr.13: 41325704-41325704 on Build GRCh38
rs746404107 Chr.13: 41325676-41325676 on Build GRCh38
rs1037825283 Chr.13: 41325602-41325603 on Build GRCh38
rs934416808 Chr.13: 41325566-41325566 on Build GRCh38
rs748698582 Chr.13: 41325694-41325694 on Build GRCh38
rs777920464 Chr.13: 41325662-41325662 on Build GRCh38
rs371851478 Chr.13: 41325665-41325665 on Build GRCh38
rs753057104 Chr.13: 41325686-41325687 on Build GRCh38
rs760662297 Chr.13: 41325739-41325739 on Build GRCh38
rs534615006 Chr.13: 41325617-41325617 on Build GRCh38
rs771978207 Chr.13: 41325714-41325714 on Build GRCh38
rs767986015 Chr.13: 41325686-41325686 on Build GRCh38
rs1011733075 Chr.13: 41325737-41325737 on Build GRCh38
rs761765263 Chr.13: 41325708-41325708 on Build GRCh38
rs774114765 Chr.13: 41325699-41325699 on Build GRCh38
rs765371802 Chr.13: 41325649-41325649 on Build GRCh38
rs919259020 Chr.13: 41325622-41325622 on Build GRCh38

View Full Product Details