Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50368193-50368418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL27
Entrez Gene ID
51264
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
L27mt
UniGene
Hs.7736
Gene Chromosome Location
Chr.17: 50367867-50373201 on Build GRCh38
Transcripts
NM_016504.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297075 c.274C>A p.L92M Chr.17: 50368265-50368265 on Build GRCh38
338202 c.333G>T p.T111T Chr.17: 50368206-50368206 on Build GRCh38
1215489 c.322C>T p.P108S Chr.17: 50368217-50368217 on Build GRCh38
1740395 c.340G>A p.V114M Chr.17: 50368199-50368199 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs939852211 Chr.17: 50368385-50368385 on Build GRCh38
rs779292144 Chr.17: 50368333-50368333 on Build GRCh38
rs764679775 Chr.17: 50368247-50368247 on Build GRCh38
rs149049206 Chr.17: 50368325-50368332 on Build GRCh38
rs58140863 Chr.17: 50368386-50368386 on Build GRCh38
rs917648086 Chr.17: 50368230-50368230 on Build GRCh38
rs147309329 Chr.17: 50368251-50368251 on Build GRCh38
rs886492183 Chr.17: 50368375-50368375 on Build GRCh38
rs1009251066 Chr.17: 50368270-50368270 on Build GRCh38
rs112763005 Chr.17: 50368338-50368341 on Build GRCh38
rs375680724 Chr.17: 50368341-50368341 on Build GRCh38
rs111380254 Chr.17: 50368314-50368321 on Build GRCh38
rs751293884 Chr.17: 50368259-50368259 on Build GRCh38
rs769906855 Chr.17: 50368227-50368227 on Build GRCh38
rs60604051 Chr.17: 50368331-50368338 on Build GRCh38
rs368243211 Chr.17: 50368228-50368228 on Build GRCh38
rs775867439 Chr.17: 50368241-50368241 on Build GRCh38
rs2586438 Chr.17: 50368338-50368338 on Build GRCh38
rs757611399 Chr.17: 50368330-50368330 on Build GRCh38
rs898424710 Chr.17: 50368392-50368392 on Build GRCh38
rs777754842 Chr.17: 50368297-50368297 on Build GRCh38
rs754769189 Chr.17: 50368272-50368272 on Build GRCh38
rs147592975 Chr.17: 50368320-50368327 on Build GRCh38
rs1039685574 Chr.17: 50368256-50368256 on Build GRCh38
rs200916450 Chr.17: 50368236-50368236 on Build GRCh38
rs372301849 Chr.17: 50368255-50368255 on Build GRCh38
rs776019884 Chr.17: 50368348-50368348 on Build GRCh38
rs778001578 Chr.17: 50368214-50368214 on Build GRCh38
rs758476657 Chr.17: 50368312-50368319 on Build GRCh38
rs747937812 Chr.17: 50368337-50368339 on Build GRCh38
rs761349230 Chr.17: 50368246-50368246 on Build GRCh38
rs759479830 Chr.17: 50368226-50368226 on Build GRCh38
rs1051875397 Chr.17: 50368371-50368371 on Build GRCh38
rs202075092 Chr.17: 50368344-50368344 on Build GRCh38
rs144076164 Chr.17: 50368240-50368240 on Build GRCh38
rs1039011080 Chr.17: 50368234-50368234 on Build GRCh38
rs767446564 Chr.17: 50368278-50368278 on Build GRCh38
rs905392218 Chr.17: 50368239-50368239 on Build GRCh38
rs199498915 Chr.17: 50368220-50368220 on Build GRCh38
rs749522448 Chr.17: 50368317-50368317 on Build GRCh38
rs535692943 Chr.17: 50368365-50368365 on Build GRCh38
rs754771906 Chr.17: 50368330-50368330 on Build GRCh38
rs751556874 Chr.17: 50368330-50368334 on Build GRCh38
rs796190278 Chr.17: 50368369-50368369 on Build GRCh38
rs144192040 Chr.17: 50368254-50368254 on Build GRCh38
rs752597675 Chr.17: 50368296-50368296 on Build GRCh38
rs968101350 Chr.17: 50368325-50368325 on Build GRCh38
rs373049296 Chr.17: 50368349-50368349 on Build GRCh38
rs781168652 Chr.17: 50368332-50368338 on Build GRCh38

View Full Product Details