Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52148356-52148617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK3
Entrez Gene ID
4752
Gene Name
NIMA related kinase 3
Gene Aliases
HSPK36
UniGene
Hs.409989
Gene Chromosome Location
Chr.13: 52132643-52159861 on Build GRCh38
Transcripts
NM_001146099.1 NM_002498.2 NM_152720.2 NR_027415.2

SNP Detail

SNP ID Chromosome Location
rs759587854 Chr.13: 52148461-52148461 on Build GRCh38
rs954655811 Chr.13: 52148397-52148397 on Build GRCh38
rs779113413 Chr.13: 52148427-52148427 on Build GRCh38
rs756648738 Chr.13: 52148404-52148404 on Build GRCh38
rs1057474487 Chr.13: 52148520-52148520 on Build GRCh38
rs762463862 Chr.13: 52148480-52148480 on Build GRCh38
rs777492498 Chr.13: 52148440-52148440 on Build GRCh38
rs772382204 Chr.13: 52148434-52148434 on Build GRCh38
rs753908743 Chr.13: 52148408-52148408 on Build GRCh38
rs746815243 Chr.13: 52148443-52148443 on Build GRCh38
rs186825318 Chr.13: 52148510-52148510 on Build GRCh38
rs754391748 Chr.13: 52148491-52148491 on Build GRCh38
rs770801231 Chr.13: 52148457-52148457 on Build GRCh38
rs192697149 Chr.13: 52148518-52148518 on Build GRCh38
rs776173649 Chr.13: 52148585-52148585 on Build GRCh38
rs1048599377 Chr.13: 52148476-52148476 on Build GRCh38
rs374088197 Chr.13: 52148465-52148465 on Build GRCh38
rs895985668 Chr.13: 52148568-52148568 on Build GRCh38
rs539178089 Chr.13: 52148458-52148458 on Build GRCh38
rs763871878 Chr.13: 52148482-52148482 on Build GRCh38
rs1046337613 Chr.13: 52148445-52148445 on Build GRCh38
rs745693180 Chr.13: 52148510-52148511 on Build GRCh38
rs200211291 Chr.13: 52148417-52148417 on Build GRCh38
rs901388964 Chr.13: 52148523-52148523 on Build GRCh38
rs762708140 Chr.13: 52148400-52148400 on Build GRCh38
rs766906334 Chr.13: 52148486-52148486 on Build GRCh38
rs553893208 Chr.13: 52148403-52148403 on Build GRCh38
rs367900950 Chr.13: 52148484-52148484 on Build GRCh38
rs573346076 Chr.13: 52148571-52148571 on Build GRCh38
rs775116270 Chr.13: 52148477-52148477 on Build GRCh38
rs182220828 Chr.13: 52148509-52148509 on Build GRCh38
rs552521187 Chr.13: 52148495-52148495 on Build GRCh38
rs571289325 Chr.13: 52148504-52148504 on Build GRCh38
rs570374527 Chr.13: 52148431-52148431 on Build GRCh38
rs780710489 Chr.13: 52148407-52148407 on Build GRCh38

View Full Product Details