Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 91127089-91127314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC69
Entrez Gene ID
100130742
Gene Name
leucine rich repeat containing 69
Gene Aliases
-
UniGene
Hs.733070
Gene Chromosome Location
Chr.8: 91102619-91219236 on Build GRCh38
Transcripts
NM_001129890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5976950 c.355C>T p.L119F Chr.8: 91127132-91127132 on Build GRCh38
6587448 c.327_348del22 p.L111fs*12 Chr.8: 91127104-91127125 on Build GRCh38
5711037 c.352C>T p.H118Y Chr.8: 91127129-91127129 on Build GRCh38
1458578 c.360G>A p.T120T Chr.8: 91127137-91127137 on Build GRCh38
4850921 c.383G>T p.R128I Chr.8: 91127160-91127160 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs577326060 Chr.8: 91127182-91127182 on Build GRCh38
rs925358878 Chr.8: 91127159-91127159 on Build GRCh38
rs771034370 Chr.8: 91127132-91127132 on Build GRCh38
rs1005851123 Chr.8: 91127270-91127270 on Build GRCh38
rs776279654 Chr.8: 91127192-91127192 on Build GRCh38
rs76359238 Chr.8: 91127125-91127125 on Build GRCh38
rs764113620 Chr.8: 91127200-91127200 on Build GRCh38
rs947724193 Chr.8: 91127173-91127173 on Build GRCh38
rs749412194 Chr.8: 91127126-91127126 on Build GRCh38
rs746143042 Chr.8: 91127172-91127172 on Build GRCh38
rs1008966161 Chr.8: 91127271-91127271 on Build GRCh38
rs991815727 Chr.8: 91127164-91127164 on Build GRCh38
rs527328654 Chr.8: 91127207-91127207 on Build GRCh38
rs998201912 Chr.8: 91127198-91127198 on Build GRCh38
rs1015843631 Chr.8: 91127282-91127282 on Build GRCh38
rs891912023 Chr.8: 91127227-91127227 on Build GRCh38
rs943429346 Chr.8: 91127163-91127163 on Build GRCh38
rs774406685 Chr.8: 91127145-91127145 on Build GRCh38
rs913116629 Chr.8: 91127124-91127124 on Build GRCh38
rs377430717 Chr.8: 91127127-91127127 on Build GRCh38
rs559814628 Chr.8: 91127196-91127196 on Build GRCh38
rs115795833 Chr.8: 91127195-91127195 on Build GRCh38
rs577944379 Chr.8: 91127201-91127201 on Build GRCh38

View Full Product Details