Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98108427-98108700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIDA
Entrez Gene ID
10247
Gene Name
reactive intermediate imine deaminase A homolog
Gene Aliases
HRSP12, P14.5, PSP, UK114
UniGene
Hs.18426
Gene Chromosome Location
Chr.8: 98102339-98117190 on Build GRCh38
Transcripts
NM_005836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273929 c.121C>A p.P41T Chr.8: 98108696-98108696 on Build GRCh38
6682186 c.157G>A p.E53K Chr.8: 98108660-98108660 on Build GRCh38
1102875 c.119A>G p.D40G Chr.8: 98108698-98108698 on Build GRCh38
6337501 c.149G>A p.G50E Chr.8: 98108668-98108668 on Build GRCh38
6336928 c.136C>G p.L46V Chr.8: 98108681-98108681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs995675832 Chr.8: 98108472-98108472 on Build GRCh38
rs757334454 Chr.8: 98108630-98108630 on Build GRCh38
rs368642363 Chr.8: 98108580-98108580 on Build GRCh38
rs150395764 Chr.8: 98108665-98108665 on Build GRCh38
rs398095992 Chr.8: 98108567-98108568 on Build GRCh38
rs967654550 Chr.8: 98108593-98108593 on Build GRCh38
rs77539382 Chr.8: 98108566-98108567 on Build GRCh38
rs928808748 Chr.8: 98108673-98108673 on Build GRCh38
rs764329287 Chr.8: 98108622-98108622 on Build GRCh38
rs898681609 Chr.8: 98108471-98108471 on Build GRCh38
rs772753535 Chr.8: 98108674-98108674 on Build GRCh38
rs753864098 Chr.8: 98108629-98108629 on Build GRCh38
rs552687035 Chr.8: 98108490-98108491 on Build GRCh38
rs1037118349 Chr.8: 98108459-98108459 on Build GRCh38
rs767978506 Chr.8: 98108597-98108597 on Build GRCh38
rs758106517 Chr.8: 98108671-98108671 on Build GRCh38
rs775552273 Chr.8: 98108605-98108605 on Build GRCh38
rs1001825853 Chr.8: 98108493-98108493 on Build GRCh38
rs75254467 Chr.8: 98108481-98108481 on Build GRCh38
rs540341734 Chr.8: 98108539-98108539 on Build GRCh38
rs760927436 Chr.8: 98108609-98108609 on Build GRCh38
rs373411556 Chr.8: 98108458-98108458 on Build GRCh38
rs1029008740 Chr.8: 98108482-98108482 on Build GRCh38
rs987875737 Chr.8: 98108648-98108648 on Build GRCh38
rs564608086 Chr.8: 98108491-98108492 on Build GRCh38
rs375649825 Chr.8: 98108535-98108535 on Build GRCh38
rs578072200 Chr.8: 98108491-98108491 on Build GRCh38
rs140448967 Chr.8: 98108664-98108664 on Build GRCh38
rs527710927 Chr.8: 98108488-98108489 on Build GRCh38
rs184746744 Chr.8: 98108585-98108585 on Build GRCh38

View Full Product Details