Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30481060-30481307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOSR1
Entrez Gene ID
9527
Gene Name
golgi SNAP receptor complex member 1
Gene Aliases
GOLIM2, GOS-28, GOS28, GOS28/P28, GS28, P28
UniGene
Hs.462680
Gene Chromosome Location
Chr.17: 30477402-30528314 on Build GRCh38
Transcripts
NM_001007024.1 NM_001007025.1 NM_004871.2 XM_005258070.4 XM_005258071.4 XM_005258072.2 XM_017025374.1 XM_017025375.1 XM_017025376.1 XM_017025377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461410 c.32-1G>C p.? Chr.17: 30481142-30481142 on Build GRCh38
6230798 c.46G>A p.A16T Chr.17: 30481157-30481157 on Build GRCh38
977309 c.77A>G p.K26R Chr.17: 30481188-30481188 on Build GRCh38
3889484 c.86C>T p.S29F Chr.17: 30481197-30481197 on Build GRCh38
977310 c.127C>T p.R43* Chr.17: 30481238-30481238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779901990 Chr.17: 30481144-30481144 on Build GRCh38
rs768484222 Chr.17: 30481262-30481262 on Build GRCh38
rs749954536 Chr.17: 30481250-30481250 on Build GRCh38
rs749222308 Chr.17: 30481260-30481260 on Build GRCh38
rs1053652856 Chr.17: 30481129-30481129 on Build GRCh38
rs200418642 Chr.17: 30481235-30481235 on Build GRCh38
rs200937047 Chr.17: 30481117-30481117 on Build GRCh38
rs780657983 Chr.17: 30481242-30481242 on Build GRCh38
rs754137941 Chr.17: 30481143-30481143 on Build GRCh38
rs765699109 Chr.17: 30481120-30481122 on Build GRCh38
rs1027852102 Chr.17: 30481119-30481119 on Build GRCh38
rs749095286 Chr.17: 30481155-30481155 on Build GRCh38
rs376535411 Chr.17: 30481090-30481090 on Build GRCh38
rs191220450 Chr.17: 30481095-30481095 on Build GRCh38
rs1007795971 Chr.17: 30481099-30481100 on Build GRCh38
rs754334701 Chr.17: 30481218-30481218 on Build GRCh38
rs756480128 Chr.17: 30481234-30481235 on Build GRCh38
rs556959471 Chr.17: 30481273-30481273 on Build GRCh38
rs760033191 Chr.17: 30481223-30481223 on Build GRCh38
rs370423863 Chr.17: 30481195-30481195 on Build GRCh38
rs756910083 Chr.17: 30481238-30481238 on Build GRCh38
rs538647813 Chr.17: 30481253-30481253 on Build GRCh38
rs772095432 Chr.17: 30481183-30481183 on Build GRCh38
rs868369585 Chr.17: 30481160-30481160 on Build GRCh38
rs760628545 Chr.17: 30481207-30481207 on Build GRCh38
rs774507380 Chr.17: 30481161-30481161 on Build GRCh38
rs766994433 Chr.17: 30481215-30481215 on Build GRCh38
rs756784314 Chr.17: 30481124-30481124 on Build GRCh38
rs965674543 Chr.17: 30481251-30481251 on Build GRCh38
rs147205774 Chr.17: 30481156-30481156 on Build GRCh38
rs878986199 Chr.17: 30481118-30481118 on Build GRCh38
rs765692418 Chr.17: 30481232-30481232 on Build GRCh38
rs768427117 Chr.17: 30481157-30481157 on Build GRCh38
rs202184797 Chr.17: 30481173-30481173 on Build GRCh38
rs755564468 Chr.17: 30481252-30481252 on Build GRCh38
rs780719843 Chr.17: 30481135-30481135 on Build GRCh38
rs377022829 Chr.17: 30481181-30481181 on Build GRCh38

View Full Product Details