Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53170330-53170601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSAD
Entrez Gene ID
51380
Gene Name
cysteine sulfinic acid decarboxylase
Gene Aliases
CSD, PCAP
UniGene
Hs.279815
Gene Chromosome Location
Chr.12: 53157663-53180909 on Build GRCh38
Transcripts
NM_001244705.1 NM_015989.4 XM_011538442.2 XM_011538446.2 XM_011538449.1 XM_011538451.1 XM_017019416.1 XM_017019417.1 XM_017019418.1 XM_017019419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5529248 c.705C>T p.V235V Chr.12: 53170446-53170446 on Build GRCh38
4043000 c.674C>T p.A225V Chr.12: 53170477-53170477 on Build GRCh38
940926 c.596C>T p.A199V Chr.12: 53170474-53170474 on Build GRCh38
4042998 c.696C>T p.T232T Chr.12: 53170455-53170455 on Build GRCh38
1586562 c.677C>T p.A226V Chr.12: 53170474-53170474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765710322 Chr.12: 53170511-53170511 on Build GRCh38
rs768237616 Chr.12: 53170494-53170494 on Build GRCh38
rs980142449 Chr.12: 53170572-53170572 on Build GRCh38
rs140047426 Chr.12: 53170363-53170363 on Build GRCh38
rs775937658 Chr.12: 53170515-53170515 on Build GRCh38
rs764103793 Chr.12: 53170521-53170521 on Build GRCh38
rs764192321 Chr.12: 53170455-53170455 on Build GRCh38
rs745745881 Chr.12: 53170478-53170478 on Build GRCh38
rs368886704 Chr.12: 53170381-53170381 on Build GRCh38
rs771814160 Chr.12: 53170507-53170507 on Build GRCh38
rs777063462 Chr.12: 53170397-53170397 on Build GRCh38
rs1040455570 Chr.12: 53170402-53170402 on Build GRCh38
rs575257751 Chr.12: 53170444-53170444 on Build GRCh38
rs1048218224 Chr.12: 53170452-53170452 on Build GRCh38
rs201933521 Chr.12: 53170474-53170474 on Build GRCh38
rs371324289 Chr.12: 53170392-53170392 on Build GRCh38
rs894329644 Chr.12: 53170571-53170571 on Build GRCh38
rs760387683 Chr.12: 53170520-53170520 on Build GRCh38
rs929313328 Chr.12: 53170451-53170451 on Build GRCh38
rs780839800 Chr.12: 53170475-53170475 on Build GRCh38
rs868842768 Chr.12: 53170445-53170445 on Build GRCh38
rs17124000 Chr.12: 53170373-53170373 on Build GRCh38
rs73099944 Chr.12: 53170364-53170364 on Build GRCh38
rs552921804 Chr.12: 53170412-53170412 on Build GRCh38
rs780023146 Chr.12: 53170550-53170550 on Build GRCh38
rs755926652 Chr.12: 53170545-53170545 on Build GRCh38
rs750278506 Chr.12: 53170416-53170417 on Build GRCh38
rs80280748 Chr.12: 53170473-53170473 on Build GRCh38
rs767824753 Chr.12: 53170543-53170543 on Build GRCh38
rs749167631 Chr.12: 53170553-53170553 on Build GRCh38
rs758520729 Chr.12: 53170450-53170450 on Build GRCh38
rs751719944 Chr.12: 53170537-53170537 on Build GRCh38
rs780191513 Chr.12: 53170486-53170486 on Build GRCh38
rs749257449 Chr.12: 53170490-53170490 on Build GRCh38
rs750165402 Chr.12: 53170544-53170544 on Build GRCh38
rs556821413 Chr.12: 53170376-53170376 on Build GRCh38
rs971404801 Chr.12: 53170549-53170549 on Build GRCh38
rs773998581 Chr.12: 53170502-53170502 on Build GRCh38
rs1016735771 Chr.12: 53170539-53170539 on Build GRCh38
rs755977061 Chr.12: 53170485-53170485 on Build GRCh38
rs375871765 Chr.12: 53170367-53170367 on Build GRCh38
rs757419251 Chr.12: 53170542-53170542 on Build GRCh38
rs747694060 Chr.12: 53170506-53170506 on Build GRCh38
rs774096318 Chr.12: 53170377-53170377 on Build GRCh38
rs767181683 Chr.12: 53170414-53170414 on Build GRCh38
rs759983714 Chr.12: 53170510-53170510 on Build GRCh38

View Full Product Details