Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39487500-39487729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT3
Entrez Gene ID
4248
Gene Name
mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase
Gene Aliases
GNT-III, GNT3
UniGene
Hs.276808
Gene Chromosome Location
Chr.22: 39447166-39492194 on Build GRCh38
Transcripts
NM_001098270.1 NM_002409.4 XM_005261608.4 XM_005261609.2 XM_006724252.2 XM_011530184.2 XM_011530185.2 XM_011530186.2 XM_011530187.2 XM_011530188.2 XM_011530189.2 XM_011530190.2 XM_011530191.2 XM_011530192.2 XM_011530194.2 XM_017028802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4104227 c.343G>A p.G115S Chr.22: 39487690-39487690 on Build GRCh38
4878119 c.270C>A p.A90A Chr.22: 39487617-39487617 on Build GRCh38
6712196 c.273G>A p.E91E Chr.22: 39487620-39487620 on Build GRCh38
4884605 c.227C>T p.S76F Chr.22: 39487574-39487574 on Build GRCh38
5755755 c.266C>T p.A89V Chr.22: 39487613-39487613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551825414 Chr.22: 39487630-39487630 on Build GRCh38
rs138382629 Chr.22: 39487641-39487641 on Build GRCh38
rs771868852 Chr.22: 39487588-39487588 on Build GRCh38
rs924665691 Chr.22: 39487576-39487576 on Build GRCh38
rs1044206604 Chr.22: 39487626-39487626 on Build GRCh38
rs796398685 Chr.22: 39487625-39487625 on Build GRCh38
rs747039606 Chr.22: 39487597-39487597 on Build GRCh38
rs761555368 Chr.22: 39487687-39487687 on Build GRCh38
rs766749026 Chr.22: 39487617-39487617 on Build GRCh38
rs776996531 Chr.22: 39487530-39487530 on Build GRCh38
rs770244016 Chr.22: 39487602-39487602 on Build GRCh38
rs148372024 Chr.22: 39487544-39487544 on Build GRCh38
rs781231522 Chr.22: 39487637-39487637 on Build GRCh38
rs369875382 Chr.22: 39487618-39487618 on Build GRCh38
rs776246246 Chr.22: 39487686-39487686 on Build GRCh38
rs778804992 Chr.22: 39487632-39487632 on Build GRCh38
rs760106779 Chr.22: 39487675-39487675 on Build GRCh38
rs200444336 Chr.22: 39487520-39487520 on Build GRCh38
rs763746572 Chr.22: 39487676-39487676 on Build GRCh38
rs62230587 Chr.22: 39487547-39487547 on Build GRCh38
rs186480026 Chr.22: 39487636-39487636 on Build GRCh38
rs562763373 Chr.22: 39487554-39487554 on Build GRCh38
rs141535501 Chr.22: 39487558-39487558 on Build GRCh38
rs200726619 Chr.22: 39487571-39487571 on Build GRCh38
rs780212481 Chr.22: 39487595-39487595 on Build GRCh38
rs199787269 Chr.22: 39487624-39487624 on Build GRCh38
rs962853713 Chr.22: 39487521-39487521 on Build GRCh38
rs773650288 Chr.22: 39487662-39487662 on Build GRCh38
rs775063824 Chr.22: 39487674-39487674 on Build GRCh38
rs200714319 Chr.22: 39487600-39487600 on Build GRCh38
rs752111231 Chr.22: 39487552-39487552 on Build GRCh38
rs745864152 Chr.22: 39487579-39487579 on Build GRCh38
rs201450879 Chr.22: 39487599-39487599 on Build GRCh38
rs368729519 Chr.22: 39487534-39487534 on Build GRCh38
rs1054415478 Chr.22: 39487612-39487612 on Build GRCh38
rs769868364 Chr.22: 39487657-39487657 on Build GRCh38
rs527409773 Chr.22: 39487623-39487623 on Build GRCh38
rs753602476 Chr.22: 39487627-39487627 on Build GRCh38
rs750142759 Chr.22: 39487633-39487633 on Build GRCh38
rs1047922656 Chr.22: 39487668-39487668 on Build GRCh38
rs757185845 Chr.22: 39487565-39487565 on Build GRCh38
rs372097244 Chr.22: 39487707-39487707 on Build GRCh38
rs763347731 Chr.22: 39487551-39487551 on Build GRCh38
rs145791006 Chr.22: 39487559-39487559 on Build GRCh38
rs750242804 Chr.22: 39487708-39487708 on Build GRCh38
rs760194893 Chr.22: 39487621-39487621 on Build GRCh38
rs763171481 Chr.22: 39487663-39487663 on Build GRCh38
rs368843548 Chr.22: 39487671-39487671 on Build GRCh38
rs773418990 Chr.22: 39487604-39487604 on Build GRCh38
rs748409695 Chr.22: 39487644-39487644 on Build GRCh38
rs762014746 Chr.22: 39487601-39487601 on Build GRCh38
rs752266312 Chr.22: 39487620-39487620 on Build GRCh38
rs867887994 Chr.22: 39487660-39487660 on Build GRCh38
rs779791255 Chr.22: 39487635-39487635 on Build GRCh38

View Full Product Details