Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33328321-33328566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK3
Entrez Gene ID
10114
Gene Name
homeodomain interacting protein kinase 3
Gene Aliases
DYRK6, FIST3, PKY, YAK1
UniGene
Hs.201918
Gene Chromosome Location
Chr.11: 33256672-33357023 on Build GRCh38
Transcripts
NM_001048200.2 NM_001278162.1 NM_001278163.1 NM_005734.4 XM_005252729.2 XM_017017076.1 XM_017017077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362752 c.1103C>T p.P368L Chr.11: 33328515-33328515 on Build GRCh38
299257 c.1098-8A>C p.? Chr.11: 33328502-33328502 on Build GRCh38
6680114 c.1101T>C p.A367A Chr.11: 33328513-33328513 on Build GRCh38
4690370 c.1128T>C p.F376F Chr.11: 33328540-33328540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768066929 Chr.11: 33328535-33328535 on Build GRCh38
rs756167571 Chr.11: 33328467-33328467 on Build GRCh38
rs752780349 Chr.11: 33328461-33328461 on Build GRCh38
rs559961611 Chr.11: 33328430-33328430 on Build GRCh38
rs749521665 Chr.11: 33328476-33328476 on Build GRCh38
rs922244111 Chr.11: 33328400-33328400 on Build GRCh38
rs973296739 Chr.11: 33328495-33328495 on Build GRCh38
rs545898827 Chr.11: 33328362-33328362 on Build GRCh38
rs982479336 Chr.11: 33328408-33328408 on Build GRCh38
rs777991429 Chr.11: 33328469-33328469 on Build GRCh38
rs775895889 Chr.11: 33328526-33328526 on Build GRCh38
rs779410675 Chr.11: 33328479-33328479 on Build GRCh38
rs746288807 Chr.11: 33328480-33328480 on Build GRCh38
rs771342395 Chr.11: 33328477-33328477 on Build GRCh38
rs776281439 Chr.11: 33328542-33328542 on Build GRCh38
rs760185882 Chr.11: 33328532-33328532 on Build GRCh38
rs776855381 Chr.11: 33328423-33328424 on Build GRCh38
rs566093690 Chr.11: 33328382-33328382 on Build GRCh38
rs772677781 Chr.11: 33328522-33328522 on Build GRCh38
rs532489486 Chr.11: 33328360-33328360 on Build GRCh38
rs980768508 Chr.11: 33328402-33328402 on Build GRCh38

View Full Product Details