Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175362602-175362869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNR
Entrez Gene ID
7143
Gene Name
tenascin R
Gene Aliases
TN-R
UniGene
Hs.659864
Gene Chromosome Location
Chr.1: 175320781-175743702 on Build GRCh38
Transcripts
NM_003285.2 XM_011509949.2 XM_017002218.1 XM_017002219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1336438 c.2810G>A p.R937Q Chr.1: 175362707-175362707 on Build GRCh38
7394544 c.2821G>T p.E941* Chr.1: 175362696-175362696 on Build GRCh38
676996 c.2809C>T p.R937W Chr.1: 175362708-175362708 on Build GRCh38
6805563 c.2736C>T p.P912P Chr.1: 175362781-175362781 on Build GRCh38
6332744 c.2721C>T p.D907D Chr.1: 175362796-175362796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766628562 Chr.1: 175362712-175362712 on Build GRCh38
rs546435742 Chr.1: 175362735-175362735 on Build GRCh38
rs191352713 Chr.1: 175362688-175362688 on Build GRCh38
rs16848290 Chr.1: 175362725-175362725 on Build GRCh38
rs61731107 Chr.1: 175362686-175362686 on Build GRCh38
rs138173384 Chr.1: 175362736-175362736 on Build GRCh38
rs755471029 Chr.1: 175362715-175362715 on Build GRCh38
rs1004611621 Chr.1: 175362711-175362711 on Build GRCh38
rs577585544 Chr.1: 175362690-175362690 on Build GRCh38
rs202100657 Chr.1: 175362841-175362841 on Build GRCh38
rs767674265 Chr.1: 175362628-175362628 on Build GRCh38
rs145986923 Chr.1: 175362691-175362691 on Build GRCh38
rs768487746 Chr.1: 175362755-175362755 on Build GRCh38
rs754407245 Chr.1: 175362714-175362714 on Build GRCh38
rs376170669 Chr.1: 175362643-175362643 on Build GRCh38
rs773416045 Chr.1: 175362707-175362707 on Build GRCh38
rs547466758 Chr.1: 175362818-175362818 on Build GRCh38
rs780674694 Chr.1: 175362642-175362642 on Build GRCh38
rs867997942 Chr.1: 175362724-175362724 on Build GRCh38
rs778676263 Chr.1: 175362677-175362677 on Build GRCh38
rs761820661 Chr.1: 175362699-175362699 on Build GRCh38
rs767727361 Chr.1: 175362704-175362704 on Build GRCh38
rs771728764 Chr.1: 175362687-175362687 on Build GRCh38
rs150401432 Chr.1: 175362708-175362708 on Build GRCh38
rs756401263 Chr.1: 175362632-175362632 on Build GRCh38
rs76938240 Chr.1: 175362756-175362756 on Build GRCh38
rs201814091 Chr.1: 175362782-175362782 on Build GRCh38
rs764028041 Chr.1: 175362823-175362823 on Build GRCh38
rs947314044 Chr.1: 175362675-175362675 on Build GRCh38
rs373577479 Chr.1: 175362625-175362625 on Build GRCh38
rs746477006 Chr.1: 175362674-175362674 on Build GRCh38
rs779685022 Chr.1: 175362750-175362750 on Build GRCh38
rs755988969 Chr.1: 175362744-175362744 on Build GRCh38
rs531733667 Chr.1: 175362729-175362729 on Build GRCh38
rs772789738 Chr.1: 175362730-175362730 on Build GRCh38
rs760776671 Chr.1: 175362786-175362786 on Build GRCh38
rs765667327 Chr.1: 175362826-175362826 on Build GRCh38
rs776882408 Chr.1: 175362813-175362813 on Build GRCh38
rs756887987 Chr.1: 175362839-175362839 on Build GRCh38
rs565454188 Chr.1: 175362648-175362648 on Build GRCh38
rs753253348 Chr.1: 175362827-175362827 on Build GRCh38
rs575878675 Chr.1: 175362808-175362808 on Build GRCh38
rs745503260 Chr.1: 175362740-175362740 on Build GRCh38
rs35262661 Chr.1: 175362647-175362648 on Build GRCh38
rs772123628 Chr.1: 175362768-175362768 on Build GRCh38
rs527422558 Chr.1: 175362803-175362803 on Build GRCh38
rs971346972 Chr.1: 175362761-175362761 on Build GRCh38
rs770042778 Chr.1: 175362635-175362635 on Build GRCh38
rs749073618 Chr.1: 175362754-175362754 on Build GRCh38
rs749161302 Chr.1: 175362689-175362689 on Build GRCh38
rs896898310 Chr.1: 175362819-175362819 on Build GRCh38
rs748291080 Chr.1: 175362759-175362759 on Build GRCh38
rs777115762 Chr.1: 175362673-175362673 on Build GRCh38
rs954939587 Chr.1: 175362633-175362633 on Build GRCh38

View Full Product Details