Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45840688-45840949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX21
Entrez Gene ID
90203
Gene Name
sorting nexin family member 21
Gene Aliases
C20orf161, PP3993, SNX-L, dJ337O18.4
UniGene
Hs.472854
Gene Chromosome Location
Chr.20: 45833418-45843275 on Build GRCh38
Transcripts
NM_001042632.2 NM_001042633.2 NM_033421.3 NM_152897.2 XM_005260608.4 XM_011529095.2 XM_011529098.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7211693 c.559C>T p.R187W Chr.20: 45840750-45840750 on Build GRCh38
6579979 c.682G>A p.E228K Chr.20: 45840873-45840873 on Build GRCh38
4501439 c.588C>T p.P196P Chr.20: 45840779-45840779 on Build GRCh38
6454957 c.590G>A p.R197H Chr.20: 45840781-45840781 on Build GRCh38
4292527 c.539G>A p.R180Q Chr.20: 45840730-45840730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757748919 Chr.20: 45840780-45840780 on Build GRCh38
rs747311073 Chr.20: 45840826-45840826 on Build GRCh38
rs761960132 Chr.20: 45840909-45840909 on Build GRCh38
rs201926135 Chr.20: 45840730-45840730 on Build GRCh38
rs533001979 Chr.20: 45840917-45840917 on Build GRCh38
rs745665174 Chr.20: 45840817-45840817 on Build GRCh38
rs759811494 Chr.20: 45840751-45840751 on Build GRCh38
rs749202684 Chr.20: 45840742-45840742 on Build GRCh38
rs752760310 Chr.20: 45840710-45840710 on Build GRCh38
rs771900758 Chr.20: 45840887-45840887 on Build GRCh38
rs753390756 Chr.20: 45840930-45840930 on Build GRCh38
rs551089787 Chr.20: 45840924-45840924 on Build GRCh38
rs956231054 Chr.20: 45840772-45840772 on Build GRCh38
rs1005332915 Chr.20: 45840858-45840858 on Build GRCh38
rs767739703 Chr.20: 45840752-45840752 on Build GRCh38
rs755797256 Chr.20: 45840857-45840857 on Build GRCh38
rs374038550 Chr.20: 45840879-45840879 on Build GRCh38
rs752448251 Chr.20: 45840852-45840852 on Build GRCh38
rs769725365 Chr.20: 45840916-45840916 on Build GRCh38
rs897018269 Chr.20: 45840889-45840889 on Build GRCh38
rs453287 Chr.20: 45840743-45840743 on Build GRCh38
rs781545083 Chr.20: 45840902-45840904 on Build GRCh38
rs151245679 Chr.20: 45840867-45840867 on Build GRCh38
rs755709413 Chr.20: 45840795-45840795 on Build GRCh38
rs763811970 Chr.20: 45840928-45840928 on Build GRCh38
rs543231945 Chr.20: 45840781-45840781 on Build GRCh38
rs201803233 Chr.20: 45840793-45840793 on Build GRCh38
rs770887895 Chr.20: 45840813-45840813 on Build GRCh38
rs35898295 Chr.20: 45840716-45840716 on Build GRCh38
rs367714719 Chr.20: 45840827-45840827 on Build GRCh38
rs370848324 Chr.20: 45840880-45840880 on Build GRCh38
rs760774166 Chr.20: 45840759-45840759 on Build GRCh38
rs766890156 Chr.20: 45840925-45840925 on Build GRCh38
rs563427177 Chr.20: 45840927-45840927 on Build GRCh38
rs764182918 Chr.20: 45840770-45840770 on Build GRCh38
rs745755088 Chr.20: 45840881-45840881 on Build GRCh38
rs760862241 Chr.20: 45840824-45840824 on Build GRCh38
rs759065882 Chr.20: 45840709-45840709 on Build GRCh38
rs935579536 Chr.20: 45840799-45840799 on Build GRCh38
rs540898085 Chr.20: 45840822-45840822 on Build GRCh38
rs370933154 Chr.20: 45840877-45840877 on Build GRCh38
rs144014614 Chr.20: 45840720-45840720 on Build GRCh38
rs758938119 Chr.20: 45840707-45840707 on Build GRCh38
rs200181120 Chr.20: 45840831-45840831 on Build GRCh38
rs140821655 Chr.20: 45840741-45840741 on Build GRCh38
rs970901595 Chr.20: 45840821-45840821 on Build GRCh38
rs905612749 Chr.20: 45840719-45840719 on Build GRCh38
rs758060801 Chr.20: 45840865-45840865 on Build GRCh38
rs751739110 Chr.20: 45840810-45840811 on Build GRCh38
rs779791518 Chr.20: 45840890-45840890 on Build GRCh38
rs757254392 Chr.20: 45840812-45840813 on Build GRCh38
rs749225472 Chr.20: 45840806-45840806 on Build GRCh38
rs200969546 Chr.20: 45840721-45840721 on Build GRCh38
rs774164764 Chr.20: 45840750-45840750 on Build GRCh38
rs777315057 Chr.20: 45840796-45840796 on Build GRCh38
rs763444966 Chr.20: 45840835-45840835 on Build GRCh38
rs776663416 Chr.20: 45840896-45840896 on Build GRCh38
rs955322898 Chr.20: 45840900-45840900 on Build GRCh38
rs766802909 Chr.20: 45840846-45840846 on Build GRCh38
rs61753670 Chr.20: 45840894-45840894 on Build GRCh38
rs777215857 Chr.20: 45840863-45840863 on Build GRCh38
rs561306037 Chr.20: 45840786-45840786 on Build GRCh38
rs190596888 Chr.20: 45840737-45840737 on Build GRCh38
rs756127266 Chr.20: 45840713-45840713 on Build GRCh38
rs546232235 Chr.20: 45840832-45840832 on Build GRCh38
rs867428701 Chr.20: 45840723-45840723 on Build GRCh38
rs374495414 Chr.20: 45840823-45840823 on Build GRCh38
rs764296889 Chr.20: 45840807-45840808 on Build GRCh38
rs747676488 Chr.20: 45840739-45840739 on Build GRCh38
rs754676255 Chr.20: 45840735-45840735 on Build GRCh38
rs557314980 Chr.20: 45840711-45840711 on Build GRCh38
rs35599678 Chr.20: 45840727-45840727 on Build GRCh38
rs778657394 Chr.20: 45840814-45840814 on Build GRCh38
rs780998892 Chr.20: 45840792-45840792 on Build GRCh38
rs775620880 Chr.20: 45840754-45840754 on Build GRCh38
rs754348584 Chr.20: 45840778-45840778 on Build GRCh38
rs768655225 Chr.20: 45840825-45840825 on Build GRCh38

View Full Product Details