Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83666301-83666536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBQLN1
Entrez Gene ID
29979
Gene Name
ubiquilin 1
Gene Aliases
DA41, DSK2, PLIC-1, UBQN, XDRP1
UniGene
Hs.9589
Gene Chromosome Location
Chr.9: 83659963-83708253 on Build GRCh38
Transcripts
NM_013438.4 NM_053067.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4524843 c.1294G>A p.E432K Chr.9: 83666388-83666388 on Build GRCh38
3659178 c.1265C>T p.P422L Chr.9: 83666417-83666417 on Build GRCh38
7113442 c.1251G>A p.M417I Chr.9: 83666431-83666431 on Build GRCh38
1288803 c.1273G>T p.A425S Chr.9: 83666409-83666409 on Build GRCh38
7113441 c.1257G>T p.L419L Chr.9: 83666425-83666425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764025915 Chr.9: 83666368-83666368 on Build GRCh38
rs964516959 Chr.9: 83666491-83666491 on Build GRCh38
rs918144410 Chr.9: 83666446-83666446 on Build GRCh38
rs199791521 Chr.9: 83666353-83666353 on Build GRCh38
rs769204636 Chr.9: 83666472-83666472 on Build GRCh38
rs765593628 Chr.9: 83666475-83666475 on Build GRCh38
rs770621039 Chr.9: 83666423-83666423 on Build GRCh38
rs372439267 Chr.9: 83666473-83666473 on Build GRCh38
rs762196173 Chr.9: 83666371-83666371 on Build GRCh38
rs772277169 Chr.9: 83666444-83666444 on Build GRCh38
rs775044665 Chr.9: 83666415-83666415 on Build GRCh38
rs954647429 Chr.9: 83666386-83666386 on Build GRCh38
rs776081346 Chr.9: 83666463-83666463 on Build GRCh38
rs760831264 Chr.9: 83666364-83666364 on Build GRCh38
rs200927958 Chr.9: 83666376-83666376 on Build GRCh38
rs141807690 Chr.9: 83666428-83666428 on Build GRCh38
rs781216899 Chr.9: 83666403-83666403 on Build GRCh38
rs140100726 Chr.9: 83666356-83666356 on Build GRCh38
rs1021805430 Chr.9: 83666352-83666352 on Build GRCh38
rs765200208 Chr.9: 83666420-83666421 on Build GRCh38
rs768054934 Chr.9: 83666358-83666358 on Build GRCh38
rs746263318 Chr.9: 83666432-83666432 on Build GRCh38
rs973328479 Chr.9: 83666495-83666495 on Build GRCh38
rs927071625 Chr.9: 83666511-83666511 on Build GRCh38
rs920539483 Chr.9: 83666507-83666507 on Build GRCh38
rs771287654 Chr.9: 83666340-83666340 on Build GRCh38
rs765546485 Chr.9: 83666374-83666374 on Build GRCh38
rs752524341 Chr.9: 83666409-83666409 on Build GRCh38
rs761024728 Chr.9: 83666464-83666464 on Build GRCh38
rs754786533 Chr.9: 83666383-83666383 on Build GRCh38
rs951256176 Chr.9: 83666338-83666338 on Build GRCh38
rs111943360 Chr.9: 83666496-83666496 on Build GRCh38
rs371738602 Chr.9: 83666479-83666479 on Build GRCh38
rs759009459 Chr.9: 83666350-83666352 on Build GRCh38
rs983223479 Chr.9: 83666517-83666517 on Build GRCh38
rs528536464 Chr.9: 83666359-83666359 on Build GRCh38
rs992814957 Chr.9: 83666441-83666441 on Build GRCh38
rs755663112 Chr.9: 83666411-83666411 on Build GRCh38
rs929238130 Chr.9: 83666510-83666510 on Build GRCh38
rs748790024 Chr.9: 83666420-83666420 on Build GRCh38

View Full Product Details