Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 234393403-234393658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARBP1
Entrez Gene ID
6894
Gene Name
TAR (HIV-1) RNA binding protein 1
Gene Aliases
TRM3, TRP-185, TRP185
UniGene
Hs.498115
Gene Chromosome Location
Chr.1: 234391313-234479174 on Build GRCh38
Transcripts
NM_005646.3 XM_005273234.3 XM_011544263.2 XM_017002195.1 XM_017002196.1 XM_017002197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554662 c.4463G>T p.G1488V Chr.1: 234393459-234393459 on Build GRCh38
7393020 c.4480G>A p.V1494I Chr.1: 234393442-234393442 on Build GRCh38
1999809 c.4494G>A p.Q1498Q Chr.1: 234393428-234393428 on Build GRCh38
905760 c.4504G>A p.D1502N Chr.1: 234393418-234393418 on Build GRCh38
6062175 c.4492C>T p.Q1498* Chr.1: 234393430-234393430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756187827 Chr.1: 234393628-234393628 on Build GRCh38
rs752785983 Chr.1: 234393605-234393605 on Build GRCh38
rs985047280 Chr.1: 234393615-234393615 on Build GRCh38
rs748937476 Chr.1: 234393499-234393499 on Build GRCh38
rs766666148 Chr.1: 234393501-234393501 on Build GRCh38
rs534784525 Chr.1: 234393445-234393445 on Build GRCh38
rs765130100 Chr.1: 234393457-234393457 on Build GRCh38
rs777401764 Chr.1: 234393494-234393495 on Build GRCh38
rs554578471 Chr.1: 234393503-234393503 on Build GRCh38
rs890579855 Chr.1: 234393437-234393437 on Build GRCh38
rs769355335 Chr.1: 234393491-234393492 on Build GRCh38
rs767620371 Chr.1: 234393530-234393530 on Build GRCh38
rs375234245 Chr.1: 234393435-234393435 on Build GRCh38
rs926963562 Chr.1: 234393520-234393520 on Build GRCh38
rs1026728642 Chr.1: 234393565-234393565 on Build GRCh38
rs771304985 Chr.1: 234393618-234393619 on Build GRCh38
rs761809322 Chr.1: 234393449-234393449 on Build GRCh38
rs146701743 Chr.1: 234393467-234393467 on Build GRCh38
rs775156497 Chr.1: 234393438-234393438 on Build GRCh38
rs994861007 Chr.1: 234393560-234393560 on Build GRCh38
rs971182827 Chr.1: 234393476-234393476 on Build GRCh38
rs930889647 Chr.1: 234393548-234393548 on Build GRCh38
rs778191185 Chr.1: 234393625-234393625 on Build GRCh38
rs1039107802 Chr.1: 234393519-234393519 on Build GRCh38
rs568423216 Chr.1: 234393571-234393571 on Build GRCh38
rs377569827 Chr.1: 234393492-234393492 on Build GRCh38
rs759628071 Chr.1: 234393524-234393524 on Build GRCh38
rs139858208 Chr.1: 234393446-234393446 on Build GRCh38
rs756610880 Chr.1: 234393622-234393622 on Build GRCh38
rs899233193 Chr.1: 234393547-234393547 on Build GRCh38
rs1051945536 Chr.1: 234393433-234393433 on Build GRCh38

View Full Product Details