Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19714600-19714849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPT5
Entrez Gene ID
5413
Gene Name
septin 5
Gene Aliases
CDCREL, CDCREL-1, CDCREL1, H5, HCDCREL-1, PNUTL1
UniGene
Hs.728762
Gene Chromosome Location
Chr.22: 19714464-19723322 on Build GRCh38
Transcripts
NM_002688.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5171155 c.39C>G p.T13T Chr.22: 19714627-19714627 on Build GRCh38
6266171 c.18G>T p.R6R Chr.22: 19714606-19714606 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747286169 Chr.22: 19714770-19714770 on Build GRCh38
rs558708440 Chr.22: 19714645-19714646 on Build GRCh38
rs770008715 Chr.22: 19714796-19714796 on Build GRCh38
rs1037792080 Chr.22: 19714809-19714809 on Build GRCh38
rs763300005 Chr.22: 19714807-19714807 on Build GRCh38
rs117051690 Chr.22: 19714751-19714751 on Build GRCh38
rs370608296 Chr.22: 19714624-19714624 on Build GRCh38
rs758068709 Chr.22: 19714831-19714831 on Build GRCh38
rs753703067 Chr.22: 19714827-19714827 on Build GRCh38
rs989898870 Chr.22: 19714712-19714712 on Build GRCh38
rs750145062 Chr.22: 19714746-19714746 on Build GRCh38
rs771302421 Chr.22: 19714626-19714626 on Build GRCh38
rs889060288 Chr.22: 19714765-19714765 on Build GRCh38
rs750285484 Chr.22: 19714816-19714816 on Build GRCh38
rs753432038 Chr.22: 19714754-19714754 on Build GRCh38
rs778591555 Chr.22: 19714755-19714755 on Build GRCh38
rs778395228 Chr.22: 19714828-19714828 on Build GRCh38
rs759749766 Chr.22: 19714777-19714777 on Build GRCh38
rs970853812 Chr.22: 19714666-19714666 on Build GRCh38
rs945453812 Chr.22: 19714729-19714729 on Build GRCh38
rs763211701 Chr.22: 19714732-19714732 on Build GRCh38
rs1025151875 Chr.22: 19714649-19714649 on Build GRCh38
rs557902022 Chr.22: 19714820-19714820 on Build GRCh38
rs752159091 Chr.22: 19714829-19714829 on Build GRCh38
rs771515751 Chr.22: 19714773-19714773 on Build GRCh38
rs761060093 Chr.22: 19714810-19714811 on Build GRCh38
rs541149811 Chr.22: 19714804-19714804 on Build GRCh38
rs376149314 Chr.22: 19714764-19714764 on Build GRCh38
rs569619443 Chr.22: 19714723-19714723 on Build GRCh38
rs375522105 Chr.22: 19714677-19714677 on Build GRCh38
rs958298664 Chr.22: 19714697-19714697 on Build GRCh38
rs767868361 Chr.22: 19714758-19714763 on Build GRCh38
rs767425384 Chr.22: 19714739-19714739 on Build GRCh38
rs993663729 Chr.22: 19714830-19714830 on Build GRCh38
rs764366773 Chr.22: 19714812-19714812 on Build GRCh38
rs765595328 Chr.22: 19714730-19714730 on Build GRCh38
rs980929028 Chr.22: 19714669-19714669 on Build GRCh38
rs773874398 Chr.22: 19714761-19714761 on Build GRCh38
rs776793256 Chr.22: 19714645-19714645 on Build GRCh38
rs922680259 Chr.22: 19714758-19714758 on Build GRCh38
rs776970540 Chr.22: 19714774-19714774 on Build GRCh38
rs371123082 Chr.22: 19714761-19714761 on Build GRCh38
rs779837958 Chr.22: 19714752-19714752 on Build GRCh38
rs921998303 Chr.22: 19714713-19714713 on Build GRCh38
rs766409895 Chr.22: 19714825-19714825 on Build GRCh38
rs185261636 Chr.22: 19714726-19714726 on Build GRCh38

View Full Product Details