Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75876250-75876505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPEF2
Entrez Gene ID
5470
Gene Name
protein phosphatase with EF-hand domain 2
Gene Aliases
PPP7CB
UniGene
Hs.290873
Gene Chromosome Location
Chr.4: 75859872-75902528 on Build GRCh38
Transcripts
NM_006239.2 XM_011532039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
294716 c.1162C>T p.R388W Chr.4: 75876445-75876445 on Build GRCh38
4839194 c.1240G>T p.E414* Chr.4: 75876367-75876367 on Build GRCh38
2152803 c.1198C>T p.R400W Chr.4: 75876409-75876409 on Build GRCh38
300911 c.1113G>T p.R371S Chr.4: 75876494-75876494 on Build GRCh38
3605882 c.1163G>A p.R388Q Chr.4: 75876444-75876444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768006854 Chr.4: 75876358-75876359 on Build GRCh38
rs753713469 Chr.4: 75876293-75876293 on Build GRCh38
rs34097437 Chr.4: 75876427-75876427 on Build GRCh38
rs149341405 Chr.4: 75876470-75876470 on Build GRCh38
rs963164692 Chr.4: 75876429-75876429 on Build GRCh38
rs140029551 Chr.4: 75876444-75876444 on Build GRCh38
rs767097910 Chr.4: 75876360-75876360 on Build GRCh38
rs947153836 Chr.4: 75876467-75876467 on Build GRCh38
rs779974619 Chr.4: 75876291-75876291 on Build GRCh38
rs756263087 Chr.4: 75876407-75876407 on Build GRCh38
rs747276411 Chr.4: 75876315-75876315 on Build GRCh38
rs896692778 Chr.4: 75876342-75876342 on Build GRCh38
rs749904288 Chr.4: 75876361-75876361 on Build GRCh38
rs749885276 Chr.4: 75876284-75876284 on Build GRCh38
rs761421627 Chr.4: 75876277-75876277 on Build GRCh38
rs774655633 Chr.4: 75876296-75876308 on Build GRCh38
rs143144324 Chr.4: 75876322-75876322 on Build GRCh38
rs376554435 Chr.4: 75876442-75876442 on Build GRCh38
rs370312706 Chr.4: 75876398-75876398 on Build GRCh38
rs1013767100 Chr.4: 75876346-75876346 on Build GRCh38
rs780202368 Chr.4: 75876457-75876457 on Build GRCh38
rs756017693 Chr.4: 75876285-75876285 on Build GRCh38
rs764349272 Chr.4: 75876270-75876270 on Build GRCh38
rs772474059 Chr.4: 75876473-75876473 on Build GRCh38
rs750694596 Chr.4: 75876453-75876453 on Build GRCh38
rs761474787 Chr.4: 75876357-75876357 on Build GRCh38
rs137955024 Chr.4: 75876435-75876435 on Build GRCh38
rs773418815 Chr.4: 75876480-75876480 on Build GRCh38
rs375630326 Chr.4: 75876296-75876296 on Build GRCh38
rs748444250 Chr.4: 75876426-75876426 on Build GRCh38
rs912418337 Chr.4: 75876274-75876274 on Build GRCh38
rs756247232 Chr.4: 75876455-75876455 on Build GRCh38
rs752494923 Chr.4: 75876379-75876379 on Build GRCh38
rs754860667 Chr.4: 75876294-75876294 on Build GRCh38
rs532974656 Chr.4: 75876312-75876312 on Build GRCh38
rs866151851 Chr.4: 75876327-75876327 on Build GRCh38
rs371148899 Chr.4: 75876428-75876428 on Build GRCh38
rs759980800 Chr.4: 75876296-75876298 on Build GRCh38
rs369913953 Chr.4: 75876320-75876320 on Build GRCh38
rs767042563 Chr.4: 75876281-75876281 on Build GRCh38
rs375702017 Chr.4: 75876461-75876461 on Build GRCh38
rs61756409 Chr.4: 75876371-75876371 on Build GRCh38
rs771240029 Chr.4: 75876318-75876318 on Build GRCh38
rs779461995 Chr.4: 75876471-75876471 on Build GRCh38
rs997482982 Chr.4: 75876403-75876403 on Build GRCh38
rs745905209 Chr.4: 75876325-75876325 on Build GRCh38
rs138339033 Chr.4: 75876481-75876481 on Build GRCh38
rs375727917 Chr.4: 75876383-75876383 on Build GRCh38
rs774710548 Chr.4: 75876417-75876417 on Build GRCh38
rs754840987 Chr.4: 75876377-75876377 on Build GRCh38
rs769044870 Chr.4: 75876352-75876352 on Build GRCh38
rs1052267231 Chr.4: 75876350-75876368 on Build GRCh38
rs765955252 Chr.4: 75876431-75876431 on Build GRCh38
rs554298761 Chr.4: 75876376-75876376 on Build GRCh38
rs774655789 Chr.4: 75876355-75876355 on Build GRCh38
rs995574336 Chr.4: 75876402-75876402 on Build GRCh38
rs776134580 Chr.4: 75876338-75876338 on Build GRCh38
rs759570831 Chr.4: 75876436-75876436 on Build GRCh38
rs369732087 Chr.4: 75876331-75876331 on Build GRCh38
rs1007263215 Chr.4: 75876388-75876388 on Build GRCh38
rs775433732 Chr.4: 75876382-75876382 on Build GRCh38
rs943871735 Chr.4: 75876280-75876280 on Build GRCh38
rs781501625 Chr.4: 75876386-75876386 on Build GRCh38
rs752548173 Chr.4: 75876438-75876438 on Build GRCh38
rs35599561 Chr.4: 75876373-75876373 on Build GRCh38
rs760808420 Chr.4: 75876478-75876479 on Build GRCh38
rs779962909 Chr.4: 75876409-75876409 on Build GRCh38
rs748640426 Chr.4: 75876472-75876472 on Build GRCh38
rs947133714 Chr.4: 75876287-75876287 on Build GRCh38
rs751257109 Chr.4: 75876271-75876271 on Build GRCh38
rs921017126 Chr.4: 75876282-75876282 on Build GRCh38
rs142757298 Chr.4: 75876412-75876412 on Build GRCh38

View Full Product Details