Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54257022-54257275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSBP3
Entrez Gene ID
23648
Gene Name
single stranded DNA binding protein 3
Gene Aliases
CSDP, SSDP, SSDP1
UniGene
Hs.476706
Gene Chromosome Location
Chr.1: 54225431-54413479 on Build GRCh38
Transcripts
NM_001009955.3 NM_018070.4 NM_145716.3 XM_006710545.3 XM_017000897.1 XM_017000898.1 XM_017000899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6790698 c.454A>G p.M152V Chr.1: 54257180-54257180 on Build GRCh38
1146347 c.461C>T p.P154L Chr.1: 54257173-54257173 on Build GRCh38
681735 c.380C>T p.P127L Chr.1: 54257173-54257173 on Build GRCh38
6790699 c.373A>G p.M125V Chr.1: 54257180-54257180 on Build GRCh38
4755532 c.452T>A p.F151Y Chr.1: 54257182-54257182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201068286 Chr.1: 54257165-54257165 on Build GRCh38
rs200322875 Chr.1: 54257103-54257103 on Build GRCh38
rs763126748 Chr.1: 54257096-54257096 on Build GRCh38
rs767938732 Chr.1: 54257110-54257110 on Build GRCh38
rs775805139 Chr.1: 54257201-54257201 on Build GRCh38
rs540126255 Chr.1: 54257223-54257223 on Build GRCh38
rs774856163 Chr.1: 54257102-54257102 on Build GRCh38
rs768789938 Chr.1: 54257206-54257206 on Build GRCh38
rs765555634 Chr.1: 54257202-54257202 on Build GRCh38
rs368799292 Chr.1: 54257170-54257170 on Build GRCh38
rs1028116856 Chr.1: 54257054-54257054 on Build GRCh38
rs761120330 Chr.1: 54257228-54257228 on Build GRCh38
rs894645525 Chr.1: 54257073-54257073 on Build GRCh38
rs955299977 Chr.1: 54257211-54257211 on Build GRCh38
rs759811105 Chr.1: 54257166-54257166 on Build GRCh38
rs752395737 Chr.1: 54257232-54257232 on Build GRCh38
rs372589100 Chr.1: 54257089-54257089 on Build GRCh38
rs374519094 Chr.1: 54257146-54257146 on Build GRCh38
rs377240726 Chr.1: 54257215-54257215 on Build GRCh38
rs372475179 Chr.1: 54257174-54257174 on Build GRCh38
rs143457918 Chr.1: 54257172-54257172 on Build GRCh38
rs757935144 Chr.1: 54257151-54257151 on Build GRCh38
rs1006253226 Chr.1: 54257149-54257149 on Build GRCh38
rs935723818 Chr.1: 54257049-54257049 on Build GRCh38
rs769868492 Chr.1: 54257199-54257199 on Build GRCh38
rs754624395 Chr.1: 54257131-54257131 on Build GRCh38
rs375387973 Chr.1: 54257230-54257230 on Build GRCh38
rs34604770 Chr.1: 54257169-54257170 on Build GRCh38
rs764459659 Chr.1: 54257098-54257098 on Build GRCh38
rs924266500 Chr.1: 54257238-54257238 on Build GRCh38
rs1054665718 Chr.1: 54257071-54257071 on Build GRCh38
rs778743195 Chr.1: 54257100-54257101 on Build GRCh38
rs746149392 Chr.1: 54257083-54257083 on Build GRCh38
rs753478248 Chr.1: 54257130-54257130 on Build GRCh38
rs773522459 Chr.1: 54257227-54257227 on Build GRCh38
rs775474722 Chr.1: 54257095-54257095 on Build GRCh38
rs781179285 Chr.1: 54257171-54257171 on Build GRCh38
rs199604326 Chr.1: 54257155-54257155 on Build GRCh38
rs1046415004 Chr.1: 54257100-54257100 on Build GRCh38
rs993921581 Chr.1: 54257104-54257104 on Build GRCh38
rs370495239 Chr.1: 54257218-54257218 on Build GRCh38
rs374408467 Chr.1: 54257099-54257099 on Build GRCh38
rs571089347 Chr.1: 54257220-54257220 on Build GRCh38
rs887889853 Chr.1: 54257118-54257118 on Build GRCh38
rs745747698 Chr.1: 54257113-54257113 on Build GRCh38
rs764970665 Chr.1: 54257145-54257145 on Build GRCh38

View Full Product Details