Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59024385-59024630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB1
Entrez Gene ID
81027
Gene Name
tubulin beta 1 class VI
Gene Aliases
-
UniGene
Hs.303023
Gene Chromosome Location
Chr.20: 59016026-59026654 on Build GRCh38
Transcripts
NM_030773.3 XM_017028085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4520824 c.1079G>A p.G360E Chr.20: 59024506-59024506 on Build GRCh38
3939419 c.1103T>C p.I368T Chr.20: 59024530-59024530 on Build GRCh38
1028550 c.1128G>T p.E376D Chr.20: 59024555-59024555 on Build GRCh38
3548258 c.1126G>A p.E376K Chr.20: 59024553-59024553 on Build GRCh38
387259 c.1161C>A p.A387A Chr.20: 59024588-59024588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774134509 Chr.20: 59024463-59024463 on Build GRCh38
rs994436900 Chr.20: 59024425-59024425 on Build GRCh38
rs200564118 Chr.20: 59024497-59024497 on Build GRCh38
rs944496829 Chr.20: 59024603-59024603 on Build GRCh38
rs780522260 Chr.20: 59024420-59024420 on Build GRCh38
rs751906254 Chr.20: 59024510-59024510 on Build GRCh38
rs374190153 Chr.20: 59024490-59024490 on Build GRCh38
rs775221566 Chr.20: 59024489-59024489 on Build GRCh38
rs567634011 Chr.20: 59024499-59024499 on Build GRCh38
rs745828581 Chr.20: 59024539-59024539 on Build GRCh38
rs866624891 Chr.20: 59024459-59024459 on Build GRCh38
rs747255971 Chr.20: 59024423-59024423 on Build GRCh38
rs777869750 Chr.20: 59024530-59024530 on Build GRCh38
rs763470972 Chr.20: 59024595-59024595 on Build GRCh38
rs781256968 Chr.20: 59024515-59024515 on Build GRCh38
rs765604740 Chr.20: 59024434-59024434 on Build GRCh38
rs199740763 Chr.20: 59024504-59024504 on Build GRCh38
rs1048726201 Chr.20: 59024439-59024439 on Build GRCh38
rs761482226 Chr.20: 59024556-59024556 on Build GRCh38
rs577942311 Chr.20: 59024542-59024542 on Build GRCh38
rs1039157941 Chr.20: 59024413-59024413 on Build GRCh38
rs770961505 Chr.20: 59024456-59024456 on Build GRCh38
rs748170810 Chr.20: 59024452-59024452 on Build GRCh38
rs748309213 Chr.20: 59024519-59024519 on Build GRCh38
rs115253190 Chr.20: 59024472-59024472 on Build GRCh38
rs763559130 Chr.20: 59024600-59024600 on Build GRCh38
rs545429143 Chr.20: 59024543-59024543 on Build GRCh38
rs766987844 Chr.20: 59024609-59024609 on Build GRCh38
rs989175665 Chr.20: 59024591-59024591 on Build GRCh38
rs759367873 Chr.20: 59024471-59024471 on Build GRCh38
rs140943896 Chr.20: 59024502-59024502 on Build GRCh38
rs556399037 Chr.20: 59024503-59024503 on Build GRCh38
rs775146221 Chr.20: 59024424-59024424 on Build GRCh38
rs148079612 Chr.20: 59024498-59024498 on Build GRCh38
rs776114449 Chr.20: 59024549-59024549 on Build GRCh38
rs751979871 Chr.20: 59024451-59024451 on Build GRCh38
rs764465342 Chr.20: 59024411-59024411 on Build GRCh38
rs755316710 Chr.20: 59024514-59024514 on Build GRCh38
rs758800483 Chr.20: 59024418-59024418 on Build GRCh38
rs763589927 Chr.20: 59024495-59024495 on Build GRCh38
rs534140732 Chr.20: 59024590-59024590 on Build GRCh38
rs1025493211 Chr.20: 59024517-59024517 on Build GRCh38
rs780577810 Chr.20: 59024506-59024506 on Build GRCh38
rs1013532966 Chr.20: 59024597-59024597 on Build GRCh38
rs764671812 Chr.20: 59024582-59024582 on Build GRCh38
rs955967093 Chr.20: 59024567-59024567 on Build GRCh38
rs773185354 Chr.20: 59024465-59024467 on Build GRCh38
rs769733428 Chr.20: 59024520-59024520 on Build GRCh38
rs781334561 Chr.20: 59024449-59024449 on Build GRCh38

View Full Product Details