Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 66369381-66369638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A26
Entrez Gene ID
115286
Gene Name
solute carrier family 25 member 26
Gene Aliases
COXPD28, SAMC
UniGene
Hs.379386
Gene Chromosome Location
Chr.3: 66133610-66380021 on Build GRCh38
Transcripts
NM_001164796.1 NM_173471.3 NR_028475.1 XM_006712956.1 XM_011533327.2 XM_017005671.1 XM_017005672.1 XM_017005673.1 XM_017005674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4158300 c.623C>T p.T208M Chr.3: 66369532-66369532 on Build GRCh38
6006975 c.572G>A p.G191E Chr.3: 66369481-66369481 on Build GRCh38
7225613 c.609A>G p.A203A Chr.3: 66369518-66369518 on Build GRCh38
4584603 c.624G>A p.T208T Chr.3: 66369533-66369533 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373100194 Chr.3: 66369594-66369594 on Build GRCh38
rs1015698482 Chr.3: 66369548-66369548 on Build GRCh38
rs2293193 Chr.3: 66369459-66369459 on Build GRCh38
rs546539897 Chr.3: 66369512-66369512 on Build GRCh38
rs112428309 Chr.3: 66369458-66369458 on Build GRCh38
rs745372502 Chr.3: 66369468-66369468 on Build GRCh38
rs768169097 Chr.3: 66369587-66369587 on Build GRCh38
rs768018446 Chr.3: 66369544-66369544 on Build GRCh38
rs779247271 Chr.3: 66369533-66369533 on Build GRCh38
rs562240562 Chr.3: 66369542-66369542 on Build GRCh38
rs776771924 Chr.3: 66369465-66369465 on Build GRCh38
rs769871437 Chr.3: 66369563-66369563 on Build GRCh38
rs751664298 Chr.3: 66369432-66369436 on Build GRCh38
rs888457942 Chr.3: 66369406-66369406 on Build GRCh38
rs1129179 Chr.3: 66369518-66369518 on Build GRCh38
rs13874 Chr.3: 66369532-66369532 on Build GRCh38
rs916233358 Chr.3: 66369608-66369608 on Build GRCh38
rs921426273 Chr.3: 66369588-66369588 on Build GRCh38
rs1016291965 Chr.3: 66369449-66369449 on Build GRCh38
rs376458397 Chr.3: 66369555-66369555 on Build GRCh38
rs372231079 Chr.3: 66369431-66369431 on Build GRCh38
rs373304440 Chr.3: 66369488-66369488 on Build GRCh38
rs368973523 Chr.3: 66369564-66369564 on Build GRCh38
rs762461496 Chr.3: 66369484-66369484 on Build GRCh38
rs529834585 Chr.3: 66369546-66369546 on Build GRCh38
rs869025315 Chr.3: 66369505-66369505 on Build GRCh38
rs762286658 Chr.3: 66369493-66369493 on Build GRCh38
rs947748447 Chr.3: 66369609-66369609 on Build GRCh38
rs761344321 Chr.3: 66369475-66369475 on Build GRCh38
rs377242376 Chr.3: 66369451-66369451 on Build GRCh38
rs775149866 Chr.3: 66369476-66369476 on Build GRCh38
rs759981319 Chr.3: 66369494-66369495 on Build GRCh38
rs748867059 Chr.3: 66369584-66369584 on Build GRCh38
rs746863887 Chr.3: 66369443-66369443 on Build GRCh38
rs757188243 Chr.3: 66369536-66369536 on Build GRCh38
rs556548845 Chr.3: 66369513-66369513 on Build GRCh38
rs932789608 Chr.3: 66369593-66369593 on Build GRCh38
rs532228857 Chr.3: 66369599-66369599 on Build GRCh38
rs577577580 Chr.3: 66369487-66369487 on Build GRCh38

View Full Product Details