Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80201744-80202017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM8
Entrez Gene ID
56978
Gene Name
PR domain 8
Gene Aliases
EPM10, PFM5
UniGene
Hs.373642
Gene Chromosome Location
Chr.4: 80183418-80204329 on Build GRCh38
Transcripts
NM_001099403.1 NM_020226.3 XM_005263144.2 XM_005263145.2 XM_005263146.4 XM_011532133.2 XM_011532135.2 XM_011532140.2 XM_011532141.2 XM_017008468.1 XM_017008469.1 XM_017008470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6755203 c.497T>C p.F166S Chr.4: 80201959-80201959 on Build GRCh38
4159228 c.511G>C p.V171L Chr.4: 80201973-80201973 on Build GRCh38
3730906 c.515C>T p.A172V Chr.4: 80201977-80201977 on Build GRCh38
5430613 c.529C>T p.R177C Chr.4: 80201991-80201991 on Build GRCh38
1619139 c.549C>G p.H183Q Chr.4: 80202011-80202011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773328126 Chr.4: 80201876-80201876 on Build GRCh38
rs770993656 Chr.4: 80201956-80201956 on Build GRCh38
rs780907674 Chr.4: 80201908-80201908 on Build GRCh38
rs374385368 Chr.4: 80201921-80201921 on Build GRCh38
rs751278347 Chr.4: 80201872-80201877 on Build GRCh38
rs199929186 Chr.4: 80201889-80201889 on Build GRCh38
rs779238085 Chr.4: 80201917-80201917 on Build GRCh38
rs981012232 Chr.4: 80201833-80201833 on Build GRCh38
rs3733334 Chr.4: 80201815-80201815 on Build GRCh38
rs144574979 Chr.4: 80201886-80201889 on Build GRCh38
rs774923926 Chr.4: 80201869-80201869 on Build GRCh38
rs141824276 Chr.4: 80201860-80201861 on Build GRCh38
rs777408932 Chr.4: 80201875-80201876 on Build GRCh38
rs957111997 Chr.4: 80201802-80201802 on Build GRCh38
rs915504819 Chr.4: 80201827-80201827 on Build GRCh38
rs772607025 Chr.4: 80201928-80201928 on Build GRCh38
rs766681984 Chr.4: 80201878-80201878 on Build GRCh38
rs908594260 Chr.4: 80201861-80201861 on Build GRCh38
rs536287220 Chr.4: 80201873-80201873 on Build GRCh38
rs755688864 Chr.4: 80201876-80201881 on Build GRCh38
rs374969203 Chr.4: 80201871-80201871 on Build GRCh38
rs756953367 Chr.4: 80201901-80201901 on Build GRCh38
rs777390514 Chr.4: 80201895-80201895 on Build GRCh38
rs758462029 Chr.4: 80201892-80201892 on Build GRCh38
rs770118482 Chr.4: 80201860-80201860 on Build GRCh38
rs56102626 Chr.4: 80201841-80201841 on Build GRCh38
rs995739627 Chr.4: 80201772-80201772 on Build GRCh38
rs554415242 Chr.4: 80201863-80201863 on Build GRCh38
rs765056822 Chr.4: 80201966-80201966 on Build GRCh38
rs773276254 Chr.4: 80201937-80201937 on Build GRCh38
rs777914988 Chr.4: 80201843-80201843 on Build GRCh38
rs573258322 Chr.4: 80201957-80201957 on Build GRCh38
rs749545671 Chr.4: 80201911-80201911 on Build GRCh38
rs753519772 Chr.4: 80201881-80201881 on Build GRCh38
rs965506424 Chr.4: 80201847-80201847 on Build GRCh38
rs1036112539 Chr.4: 80201885-80201885 on Build GRCh38
rs139033871 Chr.4: 80201775-80201775 on Build GRCh38
rs185203507 Chr.4: 80201791-80201791 on Build GRCh38
rs368993952 Chr.4: 80201872-80201872 on Build GRCh38
rs778920126 Chr.4: 80201874-80201875 on Build GRCh38
rs762974864 Chr.4: 80201976-80201976 on Build GRCh38
rs752639357 Chr.4: 80201876-80201877 on Build GRCh38
rs751186666 Chr.4: 80201899-80201899 on Build GRCh38
rs763143931 Chr.4: 80201867-80201868 on Build GRCh38
rs762625465 Chr.4: 80201886-80201887 on Build GRCh38
rs978647534 Chr.4: 80201867-80201867 on Build GRCh38
rs373009240 Chr.4: 80201885-80201888 on Build GRCh38
rs200207619 Chr.4: 80201874-80201877 on Build GRCh38
rs905357461 Chr.4: 80201953-80201953 on Build GRCh38
rs764189007 Chr.4: 80201998-80201998 on Build GRCh38
rs553329061 Chr.4: 80201877-80201877 on Build GRCh38
rs766226555 Chr.4: 80201870-80201877 on Build GRCh38
rs780519463 Chr.4: 80201886-80201893 on Build GRCh38
rs775462847 Chr.4: 80201968-80201968 on Build GRCh38
rs567356326 Chr.4: 80201860-80201863 on Build GRCh38
rs975621450 Chr.4: 80201858-80201859 on Build GRCh38
rs968251895 Chr.4: 80201832-80201832 on Build GRCh38
rs767375528 Chr.4: 80201873-80201874 on Build GRCh38
rs747105227 Chr.4: 80201948-80201948 on Build GRCh38
rs200976264 Chr.4: 80201866-80201866 on Build GRCh38
rs947247708 Chr.4: 80201862-80201862 on Build GRCh38
rs759381330 Chr.4: 80201882-80201882 on Build GRCh38
rs769094273 Chr.4: 80201914-80201914 on Build GRCh38

View Full Product Details