Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 112061148-112061377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WISP3
Entrez Gene ID
8838
Gene Name
WNT1 inducible signaling pathway protein 3
Gene Aliases
CCN6, LIBC, PPAC, PPD, WISP-3
UniGene
Hs.558428
Gene Chromosome Location
Chr.6: 112052813-112069686 on Build GRCh38
Transcripts
NM_003880.3 NM_198239.1 NR_125353.1 NR_125354.1 XM_011536220.1 XM_011536222.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
739612 c.294G>T p.K98N Chr.6: 112061182-112061182 on Build GRCh38
6902512 c.298C>T p.P100S Chr.6: 112061186-112061186 on Build GRCh38
739613 c.260G>A p.R87K Chr.6: 112061148-112061148 on Build GRCh38
1439860 c.306A>C p.E102D Chr.6: 112061194-112061194 on Build GRCh38
1672688 c.316G>T p.E106* Chr.6: 112061204-112061204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782804251 Chr.6: 112061270-112061270 on Build GRCh38
rs782644203 Chr.6: 112061315-112061315 on Build GRCh38
rs200600329 Chr.6: 112061335-112061335 on Build GRCh38
rs782667474 Chr.6: 112061206-112061206 on Build GRCh38
rs117127535 Chr.6: 112061353-112061353 on Build GRCh38
rs782261028 Chr.6: 112061233-112061233 on Build GRCh38
rs782373024 Chr.6: 112061243-112061243 on Build GRCh38
rs781834362 Chr.6: 112061293-112061293 on Build GRCh38
rs797044438 Chr.6: 112061188-112061188 on Build GRCh38
rs781906000 Chr.6: 112061333-112061333 on Build GRCh38
rs112665393 Chr.6: 112061179-112061179 on Build GRCh38
rs782099513 Chr.6: 112061265-112061265 on Build GRCh38
rs782586907 Chr.6: 112061346-112061346 on Build GRCh38
rs782468855 Chr.6: 112061185-112061185 on Build GRCh38
rs782066946 Chr.6: 112061288-112061288 on Build GRCh38
rs781928388 Chr.6: 112061257-112061257 on Build GRCh38
rs372770731 Chr.6: 112061231-112061231 on Build GRCh38
rs782191307 Chr.6: 112061323-112061324 on Build GRCh38
rs121908902 Chr.6: 112061174-112061174 on Build GRCh38
rs144622585 Chr.6: 112061256-112061256 on Build GRCh38
rs118185090 Chr.6: 112061308-112061308 on Build GRCh38
rs145747429 Chr.6: 112061269-112061269 on Build GRCh38
rs587681232 Chr.6: 112061196-112061196 on Build GRCh38
rs78266262 Chr.6: 112061252-112061252 on Build GRCh38
rs80077386 Chr.6: 112061348-112061348 on Build GRCh38
rs782446012 Chr.6: 112061221-112061221 on Build GRCh38
rs201942764 Chr.6: 112061188-112061188 on Build GRCh38
rs782025803 Chr.6: 112061338-112061338 on Build GRCh38
rs782335871 Chr.6: 112061337-112061338 on Build GRCh38
rs782346862 Chr.6: 112061267-112061267 on Build GRCh38
rs781884863 Chr.6: 112061280-112061280 on Build GRCh38
rs781995024 Chr.6: 112061268-112061268 on Build GRCh38
rs782037350 Chr.6: 112061251-112061251 on Build GRCh38
rs147337485 Chr.6: 112061190-112061190 on Build GRCh38
rs782192590 Chr.6: 112061214-112061214 on Build GRCh38

View Full Product Details