Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247589034-247589299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2G2
Entrez Gene ID
81470
Gene Name
olfactory receptor family 2 subfamily G member 2
Gene Aliases
OR1-32
UniGene
Hs.690208
Gene Chromosome Location
Chr.1: 247588360-247589313 on Build GRCh38
Transcripts
NM_001001915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348317 c.796G>T p.A266S Chr.1: 247589155-247589155 on Build GRCh38
6874546 c.676C>T p.H226Y Chr.1: 247589035-247589035 on Build GRCh38
313558 c.714G>T p.Q238H Chr.1: 247589073-247589073 on Build GRCh38
4709770 c.801G>A p.K267K Chr.1: 247589160-247589160 on Build GRCh38
7089805 c.725G>A p.G242E Chr.1: 247589084-247589084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764793959 Chr.1: 247589110-247589110 on Build GRCh38
rs763386328 Chr.1: 247589263-247589263 on Build GRCh38
rs754236597 Chr.1: 247589210-247589210 on Build GRCh38
rs371732298 Chr.1: 247589120-247589120 on Build GRCh38
rs765219287 Chr.1: 247589082-247589082 on Build GRCh38
rs772134729 Chr.1: 247589221-247589221 on Build GRCh38
rs756487359 Chr.1: 247589109-247589109 on Build GRCh38
rs766309780 Chr.1: 247589175-247589175 on Build GRCh38
rs749769720 Chr.1: 247589111-247589111 on Build GRCh38
rs376766612 Chr.1: 247589107-247589107 on Build GRCh38
rs780891464 Chr.1: 247589106-247589106 on Build GRCh38
rs910829928 Chr.1: 247589149-247589149 on Build GRCh38
rs535355641 Chr.1: 247589083-247589083 on Build GRCh38
rs200950288 Chr.1: 247589238-247589238 on Build GRCh38
rs908277676 Chr.1: 247589072-247589072 on Build GRCh38
rs767288630 Chr.1: 247589184-247589184 on Build GRCh38
rs761612257 Chr.1: 247589135-247589135 on Build GRCh38
rs770609388 Chr.1: 247589098-247589098 on Build GRCh38
rs770830134 Chr.1: 247589249-247589249 on Build GRCh38
rs759070345 Chr.1: 247589181-247589181 on Build GRCh38
rs780262118 Chr.1: 247589224-247589224 on Build GRCh38
rs150355891 Chr.1: 247589200-247589200 on Build GRCh38
rs773839582 Chr.1: 247589060-247589060 on Build GRCh38
rs751096514 Chr.1: 247589180-247589180 on Build GRCh38
rs751057296 Chr.1: 247589088-247589088 on Build GRCh38
rs775678329 Chr.1: 247589132-247589132 on Build GRCh38
rs1013204332 Chr.1: 247589252-247589252 on Build GRCh38
rs752147426 Chr.1: 247589142-247589142 on Build GRCh38
rs776452942 Chr.1: 247589071-247589071 on Build GRCh38
rs771061644 Chr.1: 247589119-247589119 on Build GRCh38
rs762751266 Chr.1: 247589165-247589165 on Build GRCh38
rs74152638 Chr.1: 247589216-247589216 on Build GRCh38
rs752329312 Chr.1: 247589193-247589193 on Build GRCh38
rs139978852 Chr.1: 247589084-247589084 on Build GRCh38
rs373547429 Chr.1: 247589070-247589070 on Build GRCh38
rs869111 Chr.1: 247589065-247589065 on Build GRCh38
rs757180401 Chr.1: 247589272-247589274 on Build GRCh38
rs149547123 Chr.1: 247589215-247589215 on Build GRCh38
rs747192851 Chr.1: 247589228-247589228 on Build GRCh38
rs769513033 Chr.1: 247589136-247589136 on Build GRCh38
rs759633082 Chr.1: 247589067-247589067 on Build GRCh38
rs765767217 Chr.1: 247589087-247589087 on Build GRCh38
rs761594043 Chr.1: 247589076-247589076 on Build GRCh38
rs756575209 Chr.1: 247589196-247589196 on Build GRCh38
rs138403077 Chr.1: 247589163-247589163 on Build GRCh38
rs745933717 Chr.1: 247589123-247589123 on Build GRCh38
rs773030936 Chr.1: 247589237-247589237 on Build GRCh38
rs754626292 Chr.1: 247589091-247589091 on Build GRCh38
rs199975925 Chr.1: 247589247-247589247 on Build GRCh38
rs769599869 Chr.1: 247589233-247589233 on Build GRCh38
rs778912851 Chr.1: 247589122-247589122 on Build GRCh38
rs865888004 Chr.1: 247589133-247589133 on Build GRCh38

View Full Product Details