Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58990861-58991134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELFCD
Entrez Gene ID
51497
Gene Name
negative elongation factor complex member C/D
Gene Aliases
HSPC130, NELF-C, NELF-D, TH1, TH1L
UniGene
Hs.517148
Gene Chromosome Location
Chr.20: 58981208-58995133 on Build GRCh38
Transcripts
NM_198976.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2764966 c.863C>T p.S288F Chr.20: 58990957-58990957 on Build GRCh38
444107 c.945G>C p.K315N Chr.20: 58991039-58991039 on Build GRCh38
2764967 c.977A>T p.E326V Chr.20: 58991071-58991071 on Build GRCh38
4755990 c.896T>G p.M299R Chr.20: 58990990-58990990 on Build GRCh38
6354548 c.981+1G>A p.? Chr.20: 58991076-58991076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769860688 Chr.20: 58991089-58991089 on Build GRCh38
rs575831844 Chr.20: 58990886-58990886 on Build GRCh38
rs776603911 Chr.20: 58990960-58990960 on Build GRCh38
rs200745856 Chr.20: 58991027-58991027 on Build GRCh38
rs369216381 Chr.20: 58990885-58990885 on Build GRCh38
rs768781929 Chr.20: 58991046-58991046 on Build GRCh38
rs375256983 Chr.20: 58990935-58990935 on Build GRCh38
rs375994512 Chr.20: 58991071-58991071 on Build GRCh38
rs61750762 Chr.20: 58990968-58990968 on Build GRCh38
rs148866352 Chr.20: 58991009-58991009 on Build GRCh38
rs761902942 Chr.20: 58990964-58990964 on Build GRCh38
rs371087478 Chr.20: 58990897-58990897 on Build GRCh38
rs759443656 Chr.20: 58991100-58991100 on Build GRCh38
rs1038705346 Chr.20: 58991059-58991059 on Build GRCh38
rs780326560 Chr.20: 58990973-58990973 on Build GRCh38
rs767283769 Chr.20: 58990983-58990983 on Build GRCh38
rs368566687 Chr.20: 58991045-58991045 on Build GRCh38
rs746243756 Chr.20: 58990934-58990934 on Build GRCh38
rs368073324 Chr.20: 58990994-58990994 on Build GRCh38
rs777121881 Chr.20: 58990892-58990892 on Build GRCh38
rs774326847 Chr.20: 58991091-58991091 on Build GRCh38
rs372830513 Chr.20: 58991066-58991066 on Build GRCh38
rs756763568 Chr.20: 58991021-58991021 on Build GRCh38
rs1037847197 Chr.20: 58991033-58991033 on Build GRCh38
rs754830320 Chr.20: 58990978-58990978 on Build GRCh38
rs199670127 Chr.20: 58990918-58990918 on Build GRCh38
rs772615957 Chr.20: 58991036-58991036 on Build GRCh38
rs746178131 Chr.20: 58990960-58990960 on Build GRCh38
rs113712592 Chr.20: 58991078-58991078 on Build GRCh38
rs375888011 Chr.20: 58991028-58991028 on Build GRCh38
rs528736476 Chr.20: 58990992-58990992 on Build GRCh38
rs163782 Chr.20: 58990888-58990888 on Build GRCh38
rs760787166 Chr.20: 58990946-58990946 on Build GRCh38
rs113288541 Chr.20: 58991076-58991076 on Build GRCh38
rs988994460 Chr.20: 58990928-58990928 on Build GRCh38
rs147821977 Chr.20: 58990919-58990919 on Build GRCh38
rs766316611 Chr.20: 58990972-58990972 on Build GRCh38
rs73297212 Chr.20: 58991107-58991107 on Build GRCh38
rs764277050 Chr.20: 58990953-58990953 on Build GRCh38
rs777436885 Chr.20: 58990999-58990999 on Build GRCh38

View Full Product Details