Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30529284-30529537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC14L6
Entrez Gene ID
730005
Gene Name
SEC14 like lipid binding 6
Gene Aliases
-
UniGene
Hs.505605
Gene Chromosome Location
Chr.22: 30522797-30546717 on Build GRCh38
Transcripts
NM_001193336.2 XM_011530357.2 XM_011530358.2 XM_011530360.2 XM_017028931.1 XM_017028932.1 XM_017028933.1 XM_017028934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3964198 c.535G>C p.E179Q Chr.22: 30529334-30529334 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747594388 Chr.22: 30529354-30529354 on Build GRCh38
rs550319423 Chr.22: 30529326-30529326 on Build GRCh38
rs570556422 Chr.22: 30529375-30529375 on Build GRCh38
rs930031423 Chr.22: 30529489-30529489 on Build GRCh38
rs56173866 Chr.22: 30529403-30529403 on Build GRCh38
rs112068496 Chr.22: 30529327-30529327 on Build GRCh38
rs762465776 Chr.22: 30529432-30529432 on Build GRCh38
rs953057224 Chr.22: 30529452-30529452 on Build GRCh38
rs1056460027 Chr.22: 30529490-30529490 on Build GRCh38
rs986157722 Chr.22: 30529465-30529465 on Build GRCh38
rs1030031890 Chr.22: 30529459-30529459 on Build GRCh38
rs900438885 Chr.22: 30529393-30529393 on Build GRCh38
rs1011832760 Chr.22: 30529359-30529359 on Build GRCh38
rs975759979 Chr.22: 30529378-30529378 on Build GRCh38
rs1017420917 Chr.22: 30529389-30529389 on Build GRCh38
rs1038459662 Chr.22: 30529486-30529486 on Build GRCh38
rs896500785 Chr.22: 30529503-30529503 on Build GRCh38
rs956421180 Chr.22: 30529353-30529353 on Build GRCh38
rs139832622 Chr.22: 30529470-30529471 on Build GRCh38
rs1053610053 Chr.22: 30529318-30529318 on Build GRCh38
rs766199638 Chr.22: 30529366-30529366 on Build GRCh38
rs7287003 Chr.22: 30529505-30529505 on Build GRCh38
rs911473165 Chr.22: 30529468-30529468 on Build GRCh38
rs373867118 Chr.22: 30529488-30529488 on Build GRCh38
rs546568941 Chr.22: 30529472-30529472 on Build GRCh38
rs865933720 Chr.22: 30529481-30529481 on Build GRCh38
rs1030759714 Chr.22: 30529337-30529337 on Build GRCh38

View Full Product Details