Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152657886-152658129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF40A
Entrez Gene ID
55660
Gene Name
pre-mRNA processing factor 40 homolog A
Gene Aliases
FBP-11, FBP11, FLAF1, FNBP3, HIP-10, HIP10, HYPA, NY-REN-6, Prp40
UniGene
Hs.643580
Gene Chromosome Location
Chr.2: 152651593-152718007 on Build GRCh38
Transcripts
NM_017892.3 XM_005246676.4 XM_005246677.4 XM_011511446.2 XM_011511447.2 XM_011511448.2 XM_011511449.2 XM_017004453.1 XM_017004454.1 XM_017004455.1 XM_017004456.1 XM_017004457.1 XM_017004458.1 XM_017004459.1 XM_017004460.1 XM_017004461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4538738 c.2596G>A p.E866K Chr.2: 152657993-152657993 on Build GRCh38
6288857 c.2299G>A p.E767K Chr.2: 152657978-152657978 on Build GRCh38
6259930 c.2355C>A p.H785Q Chr.2: 152657922-152657922 on Build GRCh38
7194190 c.2586T>A p.D862E Chr.2: 152658003-152658003 on Build GRCh38
4085673 c.2352A>G p.K784K Chr.2: 152657925-152657925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005581859 Chr.2: 152658095-152658095 on Build GRCh38
rs34255712 Chr.2: 152658043-152658044 on Build GRCh38
rs745739777 Chr.2: 152658041-152658041 on Build GRCh38
rs6713779 Chr.2: 152658069-152658069 on Build GRCh38
rs763190397 Chr.2: 152658018-152658018 on Build GRCh38
rs752141828 Chr.2: 152657922-152657922 on Build GRCh38
rs891857444 Chr.2: 152658092-152658092 on Build GRCh38
rs372511875 Chr.2: 152657982-152657982 on Build GRCh38
rs763710048 Chr.2: 152657968-152657968 on Build GRCh38
rs747248096 Chr.2: 152658005-152658005 on Build GRCh38
rs376430771 Chr.2: 152658064-152658064 on Build GRCh38
rs750858589 Chr.2: 152657985-152657985 on Build GRCh38
rs370180400 Chr.2: 152658063-152658063 on Build GRCh38
rs765336931 Chr.2: 152657917-152657917 on Build GRCh38
rs12999331 Chr.2: 152658096-152658096 on Build GRCh38
rs373293140 Chr.2: 152658027-152658027 on Build GRCh38
rs753973232 Chr.2: 152657976-152657978 on Build GRCh38
rs771766437 Chr.2: 152658045-152658045 on Build GRCh38
rs369227878 Chr.2: 152657953-152657953 on Build GRCh38
rs780259538 Chr.2: 152657995-152657995 on Build GRCh38
rs757879185 Chr.2: 152657978-152657978 on Build GRCh38
rs375449530 Chr.2: 152657992-152657992 on Build GRCh38
rs148892073 Chr.2: 152658006-152658006 on Build GRCh38
rs947956836 Chr.2: 152658053-152658053 on Build GRCh38
rs1042501567 Chr.2: 152657932-152657932 on Build GRCh38
rs760194646 Chr.2: 152657959-152657959 on Build GRCh38
rs774001324 Chr.2: 152658036-152658036 on Build GRCh38
rs781284816 Chr.2: 152658023-152658023 on Build GRCh38
rs1057158333 Chr.2: 152658073-152658073 on Build GRCh38
rs775148834 Chr.2: 152658055-152658055 on Build GRCh38
rs754346934 Chr.2: 152657969-152657969 on Build GRCh38
rs573175696 Chr.2: 152658097-152658097 on Build GRCh38
rs770947161 Chr.2: 152658030-152658030 on Build GRCh38
rs763656962 Chr.2: 152658067-152658067 on Build GRCh38

View Full Product Details