Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45499884-45500133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPM1N
Entrez Gene ID
147699
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
Gene Aliases
-
UniGene
Hs.532872
Gene Chromosome Location
Chr.19: 45497964-45502694 on Build GRCh38
Transcripts
NM_001080401.1 XM_011526474.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1394623 c.941G>A p.G314D Chr.19: 45499950-45499950 on Build GRCh38
3104297 c.986C>A p.A329D Chr.19: 45499995-45499995 on Build GRCh38
221428 c.1033G>T p.A345S Chr.19: 45500042-45500042 on Build GRCh38
6003671 c.1033G>A p.A345T Chr.19: 45500042-45500042 on Build GRCh38
3404383 c.940-1G>C p.? Chr.19: 45499948-45499948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35965696 Chr.19: 45500001-45500001 on Build GRCh38
rs763414460 Chr.19: 45499941-45499941 on Build GRCh38
rs754712804 Chr.19: 45500069-45500069 on Build GRCh38
rs547238839 Chr.19: 45499991-45499991 on Build GRCh38
rs771336269 Chr.19: 45500088-45500088 on Build GRCh38
rs143915607 Chr.19: 45499988-45499988 on Build GRCh38
rs780804243 Chr.19: 45500084-45500084 on Build GRCh38
rs564838592 Chr.19: 45499940-45499940 on Build GRCh38
rs776285397 Chr.19: 45499927-45499927 on Build GRCh38
rs529973493 Chr.19: 45500030-45500030 on Build GRCh38
rs766505464 Chr.19: 45499965-45499965 on Build GRCh38
rs909177690 Chr.19: 45499979-45499979 on Build GRCh38
rs746491040 Chr.19: 45499912-45499912 on Build GRCh38
rs943232398 Chr.19: 45499914-45499914 on Build GRCh38
rs906099926 Chr.19: 45500034-45500034 on Build GRCh38
rs768227393 Chr.19: 45499922-45499922 on Build GRCh38
rs767514876 Chr.19: 45499996-45499996 on Build GRCh38
rs774692684 Chr.19: 45499969-45499969 on Build GRCh38
rs760175067 Chr.19: 45499959-45499961 on Build GRCh38
rs770448888 Chr.19: 45499936-45499936 on Build GRCh38
rs765201209 Chr.19: 45500042-45500042 on Build GRCh38
rs779731684 Chr.19: 45500063-45500063 on Build GRCh38
rs746610280 Chr.19: 45500066-45500066 on Build GRCh38
rs758274631 Chr.19: 45500053-45500053 on Build GRCh38
rs1056300933 Chr.19: 45500033-45500033 on Build GRCh38
rs61741081 Chr.19: 45500015-45500015 on Build GRCh38
rs750183752 Chr.19: 45500051-45500051 on Build GRCh38
rs919539836 Chr.19: 45499903-45499903 on Build GRCh38
rs1011573846 Chr.19: 45500020-45500020 on Build GRCh38
rs748627241 Chr.19: 45499934-45499934 on Build GRCh38
rs746053202 Chr.19: 45500056-45500056 on Build GRCh38
rs1003404275 Chr.19: 45500023-45500023 on Build GRCh38
rs868402305 Chr.19: 45499917-45499917 on Build GRCh38

View Full Product Details